Skip to Content
Merck
All Photos(1)

Key Documents

SHC004H

Sigma-Aldrich

MISSION® pLKO.1-puro TurboGFP shRNA Control Transduction Particles, High Titer

High Titer, shRNA sequence targeting tGFP

Synonym(s):

MISSION®, MISSION® Control Transduction Particles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

product line

MISSION®

concentration

≥1x109 VP/ml (via p24 assay)

technique(s)

capture ELISA: 109 TU/mL using p24

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

The MISSION® TurboGFP shRNA Control Transduction Particles contain an shRNA sequence targeting TurboGFP. The TurboGFP shRNA Control Particles are useful as a positive knockdown control in experiments using the MISSION TurboGFP positive control vector or in cell lines expressing TurboGFP. TurboGFP is an improved variant of the green fluorescent protein copGFP cloned from copepoda Pontellina plumata.

The TurboGFP shRNA control transduction particles are produced from the sequence-verified lentiviral plasmid, pLKO.1-puro-TurboGFP shRNA (SHC004). Self-inactivating replication incompetent viral particles are produced in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids. In addition, the Control Transduction Particles are pseudotyped with an envelope G glycoprotein from Vesicular Stomatitis Virus (VSV-G), allowing transduction of a wide variety of mammalian cells. 200 μl of 109 TU/ml (via p24 titering assay) lentiviral particles are provided as frozen stock.

This high titer offering is recommended for customers: who need to utilize smaller volumes of virus, who have very difficult to transduce cell lines, or who need to transduce large numbers of cells.
When conducting experiments using MISSION® shRNA clones, the proper controls should be a key element of your experimental design to allow for accurate interpretation of knockdown results. The MISSION Control Transduction Particles are a critical positive control to monitor transduction efficiency.
To see more application data, protocols, vector maps visit sigma.com/shrna.

Application

To see more application data, protocols, vector maps visit sigma.com/shrna.

Components

High titer controls are provided as 10 x 20 μl aliquots.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
TurboGFP is a trademark of Evrogen Co.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service