Skip to Content
Merck
All Photos(1)

Key Documents

SHCLNG

Sigma-Aldrich

MISSION® shRNA

Bacterial Stock

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609

Quality Level

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

Our comprehensive shRNA product offering consists of over 150,000 pre-cloned shRNA constructs targeting more than 15,000 human and 15,000 mouse genes. The library is available in: gene family sets: gene collections related to specific functional classes such as kinases and ion channels; gene taget sets, sets of shRNA clones that target your favorite gene; and individual shRNA clones. Our shRNA and lentiviral manufacturing platforms utilize dedicated laboratories featuring state-of-the-art robotics, liquid handling and laboratory information management systems (LIMS) to provide superior quality and service for your RNAi research needs.
MISSION® TRC shRNA bacterial glycerol stocks offer a renewable resource for shRNA plasmids. These plasmids can then be transfected directly, or packaged into lentiviral particles.

Application

MISSION® shRNA has been used in the generation of damage-regulated autophagy modulator (DRAM) and p73 knockdown cell lines. It has also been used in lentivirus generation and gene knockdown and to investigate the contribution of PAX8 (paired box 8) in ovarian cancer progression.

Other Notes

To see protocols and application data please visit sigma.com/shrna.

Legal Information

Use of this product is subject to one or more license agreements.
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evidence of an oncogenic role of aberrant TOX activation in cutaneous T-cell lymphoma
Huang Y, et al.
Blood, 125(9), 1435-1443 (2015)
Nicola Currey et al.
Cellular and molecular gastroenterology and hepatology, 7(4), 819-839 (2019-03-05)
The early events by which inflammation promotes cancer are still not fully defined. The MCC gene is silenced by promoter methylation in colitis-associated and sporadic colon tumors, but its functional significance in precancerous lesions or polyps is not known. Here
A role for PAX8 in the tumorigenic phenotype of ovarian cancer cells
Di P, et al.
BMC Cancer, 14(1), 292-292 (2014)
Inhibition of damage-regulated autophagy modulator-1 (DRAM-1) impairs neutrophil differentiation of NB4 APL cells
Humbert M, et al.
Leukemia Research, 36(12), 1552-1556 (2012)
Namrata Bora-Singhal et al.
Scientific reports, 10(1), 4722-4722 (2020-03-15)
Non-small cell lung cancer (NSCLC) is known to have poor patient outcomes due to development of resistance to chemotherapy agents and the EGFR inhibitors, which results in recurrence of highly aggressive lung tumors. Even with recent success in immunotherapy using

Articles

MISSION shRNA reduce the expression of specific target genes by targeting the specific mRNA therefore reducing the corresponding protein expression.

MISSION shRNA reduce the expression of specific target genes by targeting the specific mRNA therefore reducing the corresponding protein expression.

MISSION shRNA reduce the expression of specific target genes by targeting the specific mRNA therefore reducing the corresponding protein expression.

MISSION shRNA reduce the expression of specific target genes by targeting the specific mRNA therefore reducing the corresponding protein expression.

See All

Related Content

Find shRNAs for Individual Genes. Over 150,000 pre-cloned shRNA constructs targeting more than 15,000 human and 15,000 mouse genes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service