Skip to Content
Merck
All Photos(1)

Key Documents

SAE0059

Sigma-Aldrich

OxyFluor

Synonym(s):

OF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.54

Quality Level

storage temp.

−20°C

Related Categories

Application

The Oxyrase® Enzyme System in OxyFluor removes dissolved oxygen effectively and efficiently. This significantly reduces or eliminates the formation of oxygen free radicals under conditions employed for fluorescent microscopy and results in brighter cell images. OxyFluor provides a protective effect for the biological material being studied and is an excellent solution for live cell imaging and cell culture applications.

Biochem/physiol Actions

OxyFluor is used at a 1:100 dilution. Lactate is a convenient substrate at 10 mM to 20 mM concentration.

Other Notes

There are no reports of Oxyrase® having an adverse effect on biological systems. Oxyrase® does not produce toxic compounds in the course of reducing oxygen to water.

Legal Information

OxyFluor is a trademark of Oxyrase, Inc.
Oxyrase is a registered trademark of Oxyrase, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boris Ferdman et al.
Optics express, 28(7), 10179-10198 (2020-04-01)
In microscopy, proper modeling of the image formation has a substantial effect on the precision and accuracy in localization experiments and facilitates the correction of aberrations in adaptive optics experiments. The observed images are subject to polarization effects, refractive index
Elias Nehme et al.
Nature methods, 17(7), 734-740 (2020-06-17)
An outstanding challenge in single-molecule localization microscopy is the accurate and precise localization of individual point emitters in three dimensions in densely labeled samples. One established approach for three-dimensional single-molecule localization is point-spread-function (PSF) engineering, in which the PSF is
Pernille Bülow et al.
Frontiers in cell and developmental biology, 9, 702020-702020 (2021-08-06)
Mitochondrial dysfunction has long been overlooked in neurodevelopmental disorders, but recent studies have provided new links to genetic forms of autism, including Rett syndrome and fragile X syndrome (FXS). Mitochondria show plasticity in morphology and function in response to neuronal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service