Skip to Content
Merck
All Photos(13)

Key Documents

SAB4504378

Sigma-Aldrich

Anti-phospho-DARPP-32 (pThr34) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 22 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr34)

Gene Information

human ... PPP1R1B(84152)

General description

PPP1R1B (protein phosphatase 1 regulatory inhibitor subunit 1B)/DARPP32 (dopamine- and cAMP-regulated phosphoprotein of molecular weight 32 kDa) encodes two transcripts which are further translated into two DARPP-32 protein isoforms, full length (fl)-DARPP-32 and truncated (t)-DARPP. It is located on human chromosome 17q12.

Immunogen

The antiserum was produced against synthesized peptide derived from human DARPP-32 around the phosphorylation site of Thr34.

Immunogen Range: 18-67

Biochem/physiol Actions

PPP1R1B (protein phosphatase 1 regulatory inhibitor subunit 1B)/DARPP32 (dopamine- and cAMP-regulated phosphoprotein of molecular weight 32 kDa) is considered as an inhibitor of phosphate-1 (PP-1) protein. This protein is associated with working memory and emotion processing.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrated gene copy number and expression microarray analysis of gastric cancer highlights potential target genes.
Myllykangas S, et al.
International Journal of Cancer. Journal International Du Cancer, 123(4), 817-825 (2008)
Variation of the gene coding for DARPP-32 (PPP1R1B) and brain connectivity during associative emotional learning.
Curcic-Blake B, et al.
Neuroimage, 59(2), 1540-1550 (2012)
Human t-DARPP is induced during striatal development.
Straccia M, et al.
Neuroscience, 333, 320-330 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service