Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500810

Sigma-Aldrich

Anti-Estrogen Receptor-α antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ER, ESR, ESR1, ESTR, ESTRA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 66 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ESR1(2099)

General description

Anti-Estrogen Receptor-α Antibody detects endogenous levels of total Estrogen Receptor-α protein.
Estrogen receptor α(ERα) is a member of the nuclear receptor superfamily. It is also known as ESR1 (estrogen receptor). It is located on human chromosome 6q25.1.

Immunogen

The antiserum was produced against synthesized peptide derived from human Estrogen Receptor-alpha.

Immunogen Range: 81-130

Application

Anti-Estrogen Receptor-α antibody has been used in immunohistochemistry and western blotting.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Estrogen receptor α(ERα) is involved in immune and inflammatory response. Aberrations in ESR1 results in Mayer-Rokitansky-Küster-Hauser syndrome that is associated with the congenital absence of the uterus and the upper two-thirds of the vagina.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ESR1 is co-expressed with closely adjacent uncharacterised genes spanning a breast cancer susceptibility locus at 6q25.1
Dunbier AK, et al.
PLoS Genetics, 7(4) (2011)
MYC Amplification as a Predictive Factor of Complete Pathologic Response to Docetaxel-based Neoadjuvant Chemotherapy for Breast Cancer
Pereira CBL, et al.
Clinical Breast Cancer, 17(3), 188-194 (2017)
Cynthia Brito Lins Pereira et al.
PloS one, 8(3), e60576-e60576 (2013-04-05)
Breast cancer is a complex disease, with heterogeneous clinical evolution. Several analyses have been performed to identify the risk factors for breast cancer progression and the patients who respond best to a specific treatment. We aimed to evaluate whether the
Prognostic and predictive significance of MYC and KRAS alterations in breast cancer from women treated with neoadjuvant chemotherapy
Pereira CB, et al.
PLoS ONE, 8(3) (2013)
17β-Estradiol Attenuates Hematoma Expansion through ERα/Sirt1/NF-?B Pathway in Hyperglycemic Intracerebral Hemorrhage Mice
Zheng Y, et al.
Stroke, 46(2), 485-485 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service