Skip to Content
Merck
All Photos(2)

Key Documents

04-1564

Sigma-Aldrich

Anti-Estrogen Receptor α Antibody, clone F3-A

ascites fluid, clone F3-A, from mouse

Synonym(s):

Estradiol receptor, Nuclear receptor subfamily 3 group A member 1, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 4*,5,6,7*/633 isoform, estrogen receptor alpha delta 6,7 isoform

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

F3-A, monoclonal

species reactivity

rat, human, mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ESR1(2099)

General description

Estrogen Receptor α (ERα) is a 65 kDa nuclear hormone receptor. This steroid hormone receptor is involved in the regulation of eukaryotic gene expression and affects cellular proliferation and differentiation in target tissues. ERα is a ligand-activated transcription factor and upon being bound by a ligand such as estrogen, a conformational change occurs that allows dimerization and binding to an estrogen response element. When bound to DNA, Estrogen Receptor α can positively or negatively regulate gene transcription. Like other steroid hormone receptors, Estrogen Receptors are intracellular proteins. Two ERs are known, ERα, classically known as ER, and ERβ. The Estrogen Receptor is believed to play an important role in the growth and development of a subset of hormone dependent human breast cancers. ERβ is expressed in prostate, ovary, and other reproductive organs. Although its steroid specificity is very similar to that of the ERα, the molecular mechanisms regulating the transcriptional activity of ERβ may be distinct from those regulating ERα.

Specificity

This antibody recognizes Estrogen Receptor α.

Immunogen

Epitope: Unknown
Linear peptide corresponding to human Estrogen Receptor α.

Application

Anti-Estrogen Receptor α Antibody, clone F3-A is a mouse monoclonal antibody for detection of Estrogen Receptor α also known as Estradiol receptor & Nuclear receptor subfamily 3 group A member 1 has been validated in WB, ICC.
Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected Estrogen Receptor α in A431 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blot in mouse ovary tissue lysate.

Western Blot Analysis: 1:1,000 dilution of this antibody detected Estrogen Receptor α on 10 µg of mouse ovary tissue lysate.

Target description

~ 67 kDa

Physical form

Unpurified
Unpurified mouse monoclonal IgG1κ preservative-free ascites.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Mouse ovary tissue lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoxiang Cheng et al.
Development, growth & differentiation, 58(3), 327-338 (2016-04-03)
During mouse early embryogenesis, blastomeres increase in number by the morula stage. Among them, the outer cells are polarized and differentiated into trophectoderm (TE), while the inner cells remain unpolarized and give rise to inner cell mass (ICM). TE provides
Effects of estrogen on vascular inflammation: a matter of timing.
Novella, S; Heras, M; Hermenegildo, C; Dantas, AP
Arteriosclerosis, Thrombosis, and Vascular Biology null
Xiaoman Dai et al.
Neuroendocrinology, 113(7), 756-769 (2023-01-04)
Gonadotropin-releasing hormone (GnRH) is a hypothalamic neuropeptide that plays important roles in the female fertility. Accumulating evidence suggests that ERα present in the astrocytes of the hypothalamus region is essential for production of GnRH. The astrocytes display age-related senescence associated
Willian O Dos Santos et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2023-08-26)
Dysfunctions in growth hormone (GH) secretion increase the prevalence of anxiety and other neuropsychiatric diseases. GH receptor (GHR) signaling in the amygdala has been associated with fear memory, a key feature of post-traumatic stress disorder. However, it is currently unknown
Vittoria Poli et al.
Nature communications, 9(1), 1024-1024 (2018-03-11)
Breast cancer consists of highly heterogeneous tumors, whose cell of origin and driver oncogenes are difficult to be uniquely defined. Here we report that MYC acts as tumor reprogramming factor in mammary epithelial cells by inducing an alternative epigenetic program

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service