Skip to Content
Merck
All Photos(1)

Key Documents

SAB4500753

Sigma-Aldrich

Anti-APOBEC3A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Phorbolin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 23 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

APOBEC3A (apolipoprotein B mRNA editing enzyme catalytic subunit 3A) belongs to the AID/APOBEC family of enzymes. It is located on human chromosome 22q13.1.
Anti-APOBEC3A antibody detects endogenous levels of total APOBEC3A protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human APOBEC3A.

Immunogen Range: 27-76

Application

Anti-APOBEC3A antibody has been used in western blotting.

Biochem/physiol Actions

APOBEC3A (apolipoprotein B mRNA editing enzyme catalytic subunit 3A) deaminates methylated cytosine bases in DNA. It is considered as a viral DNA mutator and performs broad antiviral activity.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heat Increases the Editing Efficiency of Human Papillomavirus E2 Gene by Inducing Upregulation of APOBEC3A and 3G
Yang Y, et al.
The Journal of Investigative Dermatology, 137(4), 810-818 (2017)
Human Tribbles 3 protects nuclear DNA from cytidine deamination by APOBEC3A
Aynaud MM, et al.
The Journal of Biological Chemistry (2012)
APOBEC3A efficiently deaminates methylated, but not TET-oxidized, cytosine bases in DNA
Schutsky EK, et al.
Nucleic Acids Research, 45(13), 7655-7665 (2017)
Jan-Hendrik Bockmann et al.
The Journal of infectious diseases, 220(4), 567-577 (2019-03-30)
Type III interferons (IFNs) (λ1-3) activate similar signaling cascades as type I IFNs (α and β) via different receptors. Since IFN-α and lymphotoxin-β activate cytosine deamination and subsequent purging of nuclear hepatitis B virus (HBV) DNA, we investigated whether IFN-β
Rodolphe Suspène et al.
Nucleic acids research, 45(6), 3231-3241 (2017-01-20)
Foreign and self-cytoplasmic DNA are recognized by numerous DNA sensor molecules leading to the production of type I interferons. Such DNA agonists should be degraded otherwise cells would be chronically stressed. Most human APOBEC3 cytidine deaminases can initiate catabolism of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service