Skip to Content
Merck
All Photos(1)

Key Documents

M8770

Sigma-Aldrich

Anti-Mouse IgG1 (heavy chain specific) antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Mouse Igg1, Heavy Chain Specific Secondary Antibody, Heavy Chain Specific Secondary Antibody - Anti-Mouse IgG1 (heavy chain specific) antibody produced in goat

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

200
300

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

technique(s)

Ouchterlony double diffusion: suitable

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin IgG is a glycoprotein antibody with two light chains and a pair of identical heavy chains.

Immunogen

Mouse IgG1 (myeloma protein)

Application

Anti-Mouse IgG1 (heavy chain specific) antibody is suitable for use in immunoelectrophoresis and Ouchterlony double diffusion. The antibody may also be used in immunoblot .
Anti-Mouse IgG1 (heavy chain specific) antibody produced in goat has been used in immunohistochemistry and enzyme-linked immunosorbent assay (ELISA)

Biochem/physiol Actions

Digestion of IgG by papain results in the generation of fragment antigen-binding (Fab). Pepsin digestion of IgG results in fragment crystallizable (fc). IgG1 is most abundant of IgG types and its deficiency results in hypogammaglobulinemia.

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2.

Reconstitution

Reconstitute with 0.135 M sodium chloride.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A H Schrijvers et al.
Cancer research, 53(18), 4383-4390 (1993-09-15)
Using viable cells of a human head and neck squamous cell carcinoma (HNSCC) cell line as immunogen, we developed monoclonal antibody (MAb) U36. Immunohistochemical examination revealed distinct surface labeling of MAb U36 with normal human squamous epithelium and squamous cell
The combined effect of mesenchymal stem cells and resveratrol on type 1 diabetic neuropathy
Wang C, et al.
Experimental and Therapeutic Medicine, 17(5), 3555-3563 (2019)
J Pass et al.
Scandinavian journal of immunology, 58(3), 298-305 (2003-09-03)
The urokinase receptor (uPAR) is a glycolipid-anchored cell surface glycoprotein that plays a central role in extracellular proteolysis during tissue remodeling processes including cancer invasion. Furthermore, uPAR is found on the surface of both dendritic cells (DCs) and T cells
Louis-P Vézina et al.
Plant biotechnology journal, 7(5), 442-455 (2009-05-09)
Plant-based transient expression is potentially the most rapid and cost-efficient system for the production of recombinant pharmaceutical proteins, but safety concerns associated with plant-specific N-glycosylation have hampered its adoption as a commercial production system. In this article, we describe an
Kai Yao et al.
Drug design, development and therapy, 13, 387-396 (2019-02-06)
As a novel antidepressant drug, agomelatine has good therapeutic effect on the mood disorder and insomnia in Alzheimer's disease (AD). Recent studies have shown the neuroprotective function of agomelatine, including anti-oxidative and anti-apoptosis effect. However, it remains unclear whether agomelatine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service