Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500437

Sigma-Aldrich

Anti-Claudin 7 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CEPTRL2, claudin 7, claudin 9, claudin-1, claudin-7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 22 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLDN7(1366)

Related Categories

General description

Anti-Claudin 7 Antibody detects endogenous levels of total Claudin 7 protein.
Claudin 7 is a tight junction membrane protein that belongs to claudins (cld) family. It is upregulated at high level in the upper gastrointestinal tract and the liver. It is located on human chromosome 17p13.

Immunogen

The antiserum was produced against synthesized peptide derived from human Claudin 7.

Immunogen Range: 162-211

Biochem/physiol Actions

In HCC827 and NCI-H358 lung cancer cells, suppression of claudin-7 decreases the expression of integrin β1. Claudins are important constituents of tight junctions (TJ) in the upper region of epithelial cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harold Love et al.
Tissue engineering. Part A, 26(19-20), 1091-1098 (2020-04-22)
Patient-oriented applications of cell culture include cell therapy of organ failure like chronic renal failure. Clinical deployment of a cell-based device for artificial renal replacement requires qualitative and quantitative fidelity of a cultured cell to its in vivo counterpart. Active
The claudins
Lal-Nag M and Morin P J
Genome Biology (2009)
A non-tight junction function of claudin-7-Interaction with integrin signaling in suppressing lung cancer cell proliferation and detachment
Lu Z, et al.
Molecular Cancer (2015)
The importance of claudin-7 palmitoylation on membrane subdomain localization and metastasis-promoting activities
Heiler S, et al.
Cell communication and signaling : CCS (2015)
Sijo Mathew et al.
Development (Cambridge, England), 144(22), 4148-4158 (2017-10-11)
Kidney collecting system development requires integrin-dependent cell-extracellular matrix interactions. Integrins are heterodimeric transmembrane receptors consisting of α and β subunits; crucial integrins in the kidney collecting system express the β1 subunit. The β1 cytoplasmic tail has two NPxY motifs that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service