Skip to Content
Merck
All Photos(1)

Key Documents

HPA004823

Sigma-Aldrich

Anti-TNC antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DFNA56, Anti-HXB, Anti-MGC167029, Anti-TN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

RLVKLIPGVEYLVSIIAMKGFEESEPVSGSFTTALDGPSGLVTANITDSEALARWQPAIATVDSYVISYTGEKVPEITRTVSGNTVEYALTDLEPATEYTLRIFAEKGPQKSSTITAKFTTDLDSPRDLTATEV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNC(3371)

General description

TNC (Tenascin C) is a large, multicellular, extracellular matrix (ECM) glycoprotein belonging to the tenascin family. Generally, it is expressed in normal adult tissue at very low level but highly expressed in tumor cells. Elevated expression level has been reported within the outflow side of the aortic valve leaflets.

Immunogen

Tenascin precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-TNC antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

TNC (Tenascin C) is involved in the tissue remodeling and apoptosis. The pleiotropic regulator,TNC, is associated with various cellular processes such as embryogenesis, wound healing, cell proliferation, differentiation, motility, and nerve regeneration. It is also associated with various cardiac and vascular diseases. It acts as a cardiac biomarker in the detection of various cardiac diseases, including heart failure, thrombosis, atherosclerosis, and cancer. It also controls the regulation of neural differentiation signaling in bone marrow-derived human mesenchymal stem cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86782

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hung-Li Tsai et al.
Tissue engineering. Part A, 20(13-14), 1908-1921 (2014-05-16)
Mesenchymal stem cells (MSCs) are currently thought to transdifferentiate into neural lineages under specific microenvironments. Studies have reported that the tenascin family members, tenascin-C (TnC) and tenascin-R (TnR), regulate differentiation and migration, in addition to neurite outgrowth and survival in
Irena Niebroj-Dobosz
Clinica chimica acta; international journal of clinical chemistry, 413(19-20), 1516-1518 (2012-06-13)
Tenascin-C (TN-C), a hexameric extracellular matrix glycoprotein, is a pleiotropic regulator of a variety of cell functions associated with embryogenesis, wound healing, cell proliferation, differentiation, motility, and nerve regeneration. Due to its role in remodeling processes, TN-C is involved with
Hidenori Suzuki et al.
Acta neurochirurgica. Supplement, 120, 117-121 (2014-11-05)
Tenascin-C (TNC), a matricellular protein, exerts diverse functions, including tissue remodeling and apoptosis, and is induced in cerebrospinal fluid (CSF) after aneurysmal subarachnoid hemorrhage (SAH). The purpose of this study was to examine the relationships among CSF TNC levels, initial
Yingqiang Xu et al.
Diagnostic pathology, 10, 211-211 (2015-12-15)
Matrix metalloproteinase-9 (MMP-9) and Tenascin-C (TN-C) have been shown to be involved in the metastasis of many tumors. The purpose of this study was to determine the relationship between the co-expression of these two molecules and the clinical prognosis of
Vijay Sharma et al.
Cureus, 16(7), e65566-e65566 (2024-08-28)
Background The use of corticosteroids, such as methylprednisolone, for pain management is a common clinical practice. However, it is well known that corticosteroids induce toxicity in anterior cruciate ligament (ACL)-derived stromal cells and chondrocytes. Growth factors from platelets have anti-inflammatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service