Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300825

Sigma-Aldrich

Anti-COL4A1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

HANAC, ICH, POREN1, arresten

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

concentration

1.1 mg/mL

technique(s)

immunohistochemistry: 1:50- 1:200

isotype

IgG

accession no.

NP_001836.2

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COL4A1(1282)

Specificity

The antibody detects endogenous levels of total COL4A1 protein.

Immunogen

Synthetic peptide corresponding to residues near the C terminal of human collagen, type IV, alpha 1

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.3 PBS, 0.05% NaN3, 50% Glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mai Kikumoto et al.
Brain communications, 5(6), fcad281-fcad281 (2023-11-13)
Pontine autosomal dominant microangiopathy and leukoencephalopathy is one of hereditary cerebral small vessel diseases caused by pathogenic variants in COL4A1 3'UTR and characterized by multiple small infarctions in the pons. We attempted to establish radiological features of this disease. We
Feng Wu et al.
Endocrine-related cancer (2019-03-15)
Tumour-derived exosomes under hypoxic conditions contain informative microRNAs involved in the interaction of cancer and para-carcinoma cells, thus contributing to tissue remodelling of the tumour microenvironment (TME). Exosomes isolated from hypoxic papillary thyroid cancer cells, BCPAP cells and KTC-1 cells
Qiang-Nu Zhang et al.
Cancer management and research, 11, 4757-4772 (2019-06-20)
Background: The systematic expression characteristics and functions of collagen genes in gastric cancer (GC) have not been reported. Through public data integration, combined with bioinformatics analysis, we identified a panel of collagen genes overexpressed in GC. The functions of these genes
Meaghan Griffiths et al.
Scientific reports, 9(1), 8644-8644 (2019-06-16)
The endometrium is a highly complex tissue that is vulnerable to subtle gene expression changes and is the first point of contact for an implanting blastocyst. Successful blastocyst implantation can only occur when the endometrium is receptive during a short
Akira Sugiyama et al.
The Journal of veterinary medical science, 81(4), 522-531 (2019-02-07)
The basement membrane surrounding cardiomyocytes is mainly composed of α1 and α2 chain of type IV collagen. Arresten and canstatin are fragments of non-collagenous C-terminal domain of α1 and α2 chain, respectively. We previously reported that the expression of canstatin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service