Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300696

Sigma-Aldrich

Anti-Postn antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Ost2, Anti-PLF, Anti-PN, Anti-peri, Anti-periostin, osteoblast specific factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~93 kDa

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(R-E-E-I-E)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Postn(50706)

General description

Periostin (POSTN) gene with 23 exons spanning 36 kb is mapped to chromosome 13q13.3. The gene codes for periostin, a matricellular protein that belongs to the family of non-structural matrix proteins. Periostin structure contains a signal peptide, four fasciclin-like domains (FAS1) with vitamin K-dependent γ-carboxyglutamic acid (Gla) residues, and hydrophilic C-terminal region with the heparin-binding site. The protein is mainly expressed in the periosteum, periodontal ligament, heart valves, and tendons. The antibody detects endogenous level of total periostin protein.

Immunogen

Peptide sequence around aa. 125-129 ( R-E-E-I-E), according to the protein NP_006466.2

Application

Anti-POSTN antibody produced in rabbit has been used in western blot.

Biochem/physiol Actions

Periostin (POSTN) facilitates extracellular matrix structure and organization. It acts as a collagen chaperone and regulates type I collagen fibrillogenesis. POSTN is implicated in early stages of bone formation, tissue repair, and cell recruitment and adhesion. This protein is essential for cancer stem cell maintenance and metastasis. Increased expression of POSTN has been observed in various types of cancers including prostate, breast, lung, pancreatic, ovarian, and colorectal cancer. Hence, the postn levels can be used as a prognostic biomarker for these cancers.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Binds to heparin. Induces cell attachment and spreading and plays a role in cell adhesion. May play a role in extracellular matrix mineralization.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Periostin Mediates Oestrogen-Induced Osteogenic Differentiation of Bone Marrow Stromal Cells in Ovariectomised Rats.
Li, et al.
BioMed Research International, 2020, 9405909-9405909 (2020)
Ilaria Malanchi et al.
Nature, 481(7379), 85-89 (2011-12-14)
Metastatic growth in distant organs is the major cause of cancer mortality. The development of metastasis is a multistage process with several rate-limiting steps. Although dissemination of tumour cells seems to be an early and frequent event, the successful initiation
Nicolas Bonnet et al.
The Journal of biological chemistry, 284(51), 35939-35950 (2009-10-20)
Periostin (gene Postn) is a secreted extracellular matrix protein involved in cell recruitment and adhesion and plays an important role in odontogenesis. In bone, periostin is preferentially expressed in the periosteum, but its functional significance remains unclear. We investigated Postn(-/-)
Laura González-González et al.
Frontiers in oncology, 8, 225-225 (2018-06-28)
Tumor microenvironment is considered nowadays as one of the main players in cancer development and progression. Tumor microenvironment is highly complex and consists of non-tumor cells (i.e., cancer-associated fibroblast, endothelial cells, or infiltrating leukocytes) and a large list of extracellular
Collagen structure, folding and function
A Text Book of Fibre Science and Technology, 71-84 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service