Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200593

Sigma-Aldrich

Anti-Occludin antibody from rabbit

affinity isolated antibody

Synonym(s):

BLCPMG, OCLN, PPP1R115

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Occludin (OCLN) is an integral membrane protein, encoded by the gene mapped to human chromosome 5q13.2. OCLN is a ~65 kDa protein with four transmembrane domains including, a long COOH-terminal cytoplasmic domain, a short NH2-terminal cytoplasmic domain, two extracellular loops, and one intracellular turn. It is specifically localized at tight junctions (TJ).

Application

Anti-Occludin antibody from rabbit has been used in immunocytochemistry and western blotting.

Biochem/physiol Actions

Occludin (OCLN), along with tight junction (TJ)-associated peripheral membrane proteins, plays a vital role in formation and regulation of TJ. The encoded protein facilitates the translocation of p85a to TJs to regulate actin organization after oxidative stress. OCLN is also implicated in cell migration. Mutation in the gene leads to the development of band-like calcification with simplified gyration and polymicrogyria (BLC-PMG).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudin-1 and -2: Novel Integral Membrane Proteins Localizing at Tight Junctions with No Sequence Similarity to Occludin.
Mikio F, et al.
The Journal of Cell Biology, 141(7), 1539?1550-1539?1550 (1998)
Splicing diversity of the human OCLN gene and its biological significance for hepatitis C virus entry.
Kohaar I, et al.
Journal of Virology, 84(14), 6987-6994 (2010)
Recessive Mutations in the Gene Encoding the Tight Junction Protein Occludin Cause Band-like Calcification with Simplified Gyration and Polymicrogyria.
Mary C O, et al.
American Journal of Human Genetics, 87(3), 354?364-354?364 (2010)
Dongli Li et al.
Stem cell research & therapy, 12(1), 24-24 (2021-01-09)
Subretinal fibrosis resulting from neovascular age-related macular degeneration (nAMD) is one of the major causes of serious and irreversible vision loss worldwide, and no definite and effective treatment exists currently. Retinal pigmented epithelium (RPE) cells are crucial in maintaining the
The tight junction protein, occludin, regulates the directional migration of epithelial cells.
Du D, et al.
Developmental Cell, 18(1), 52-63 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service