Skip to Content
Merck
All Photos(2)

Key Documents

AV41490

Sigma-Aldrich

Anti-FN1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Fn1 Antibody, Fn1 Antibody - Anti-FN1 antibody produced in rabbit, Anti-CIG, Anti-DKFZp686F10164, Anti-DKFZp686H0342, Anti-DKFZp686I1370, Anti-DKFZp686O13149, Anti-FINC, Anti-FN, Anti-Fibronectin 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

76 kDa

species reactivity

bovine, dog, sheep, pig, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FN1(2335)

General description

Fibronectins are a class of immunochemically related glycoproteins present in basement membranes, collective tissues and blood wherein they mediate adhesion between matrix components and cells. Plasma fibronectin (CLG) mediates the attachment of monocytes (fibroblasts, macrophages) to various cell matrices and materials such as gelatin.

Specificity

Anti-FN1 polyclonal antibody reacts with human, canine, rabbit, bovine, rat, and mouse plasma fibronectin (CIG).

Immunogen

Synthetic peptide directed towards the C terminal region of human FN1

Application

Anti-FN1 polyclonal antibody is used to tag plasma fibronectin (CIG) for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the possible roles of fibronectin (CIG) in the adherence of monocytes to cell matricies and solid surfaces coated with materials such as gelatin.

Biochem/physiol Actions

FN1 is a glycoprotein present in a soluble dimeric form in plasma, and in a dimeric or multimeric form at the cell surface and in extracellular matrix. Fibronectin is involved in cell adhesion and migration processes including embryogenesis, wound healing, blood coagulation, host defense, and metastasis.

Sequence

Synthetic peptide located within the following region: NCRRPGGEPSPEGTTGQSYNQYSQRYHQRTNTNVNCPIECFMPLDVQADR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Simone Buchtler et al.
Journal of the American Society of Nephrology : JASN, 29(7), 1859-1873 (2018-05-20)
Background Interstitial fibrosis is associated with chronic renal failure. In addition to fibroblasts, bone marrow-derived cells and tubular epithelial cells have the capacity to produce collagen. However, the amount of collagen produced by each of these cell types and the
Saidou Balam et al.
Frontiers in immunology, 12, 816509-816509 (2022-02-08)
Fibrosis is a prominent feature of chronic allograft rejection, caused by an excessive production of matrix proteins, including collagen-1. Several cell types produce collagen-1, including mesenchymal fibroblasts and cells of hematopoietic origin. Here, we sought to determine whether tissue-resident donor-derived
Guiqin Song et al.
Oncotarget, 8(11), 17771-17784 (2017-02-02)
Esophageal cancer is a highly aggressive malignancy with very poor overall prognosis. Given the strong clinical relevance of SATB1 in esophagus cancer and other cancers suggested by previous studies, the exact function of SATB1 in esophagus cancer development is still
Saidou Balam et al.
Journal of immunology (Baltimore, Md. : 1950), 202(12), 3514-3523 (2019-05-10)
Chronic rejection is a major problem in transplantation medicine, largely resistant to therapy, and poorly understood. We have shown previously that basophil-derived IL-4 contributes to fibrosis and vasculopathy in a model of heart transplantation with depletion of CD4+ T cells.
Xianghui Chen et al.
Kidney international, 95(4), 880-895 (2019-02-23)
Ectopic fat deposition (EFD) in the kidney has been shown to play a causal role in diabetic nephropathy; however, the mechanism underlying EFD remains elusive. By transcriptome analysis, we found decreased expression levels of disulfide-bond A oxidoreductase-like protein (DsbA-L) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service