Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500794

Sigma-Aldrich

Anti-PINK1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-BRPK, Anti-FLJ27236, Anti-PARK6, Anti-PTEN induced putative kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
indirect ELISA: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PINK1(65018)

Immunogen

Peptide with sequence C-QGKAHLESRSYQEAQ from the internal region of the protein sequence according to NP_115785.1.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc Beltrà et al.
Nature communications, 14(1), 1849-1849 (2023-04-04)
Cachexia is a debilitating wasting syndrome and highly prevalent comorbidity in cancer patients. It manifests especially with energy and mitochondrial metabolism aberrations that promote tissue wasting. We recently identified nicotinamide adenine dinucleotide (NAD+) loss to associate with muscle mitochondrial dysfunction
Riccardo Ballarò et al.
Cancers, 13(4) (2021-03-07)
Objective: Cachexia is a complex metabolic syndrome frequently occurring in cancer patients and exacerbated by chemotherapy. In skeletal muscle of cancer hosts, reduced oxidative capacity and low intracellular ATP resulting from abnormal mitochondrial function were described. Methods: The present study
Zaira Aversa et al.
Scientific reports, 6, 30340-30340 (2016-07-28)
Basal rates of autophagy can be markedly accelerated by environmental stresses. Recently, autophagy has been involved in cancer-induced muscle wasting. Aim of this study has been to evaluate if autophagy is induced in the skeletal muscle of cancer patients. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service