Skip to Content
Merck
All Photos(1)

Key Documents

SAB1406869

Sigma-Aldrich

Anti-PUM1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

HSPUM, KIAA0099, PUMH, PUMH1, PUML1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~126.5 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PUM1(9698)

General description

This gene encodes a member of the PUF family, evolutionarily conserved RNA-binding proteins related to the Pumilio proteins of Drosophila and the fem-3 mRNA binding factor proteins of C. elegans. The encoded protein contains a sequence-specific RNA binding domain comprised of eight repeats and N- and C-terminal flanking regions, and serves as a translational regulator of specific mRNAs by binding to their 3′ untranslated regions. The evolutionarily conserved function of the encoded protein in invertebrates and lower vertebrates suggests that the human protein may be involved in translational regulation of embryogenesis, and cell development and differentiation. Alternatively spliced transcript variants encoding different isoforms have been described. (provided by RefSeq)

Immunogen

PUM1 (NP_055491, 1 a.a. ~ 1186 a.a) full-length human protein.

Sequence
MSVACVLKRKAVLWQDSFSPHLKHHPQEPANPNMPVVLTSGTGSQAQPQPAANQALAAGTHSSPVPGSIGVAGRSQDDAMVDYFFQRQHGEQLGGGGSGGGGYNNSKHRWPTGDNIHAEHQVRSMDELNHDFQALALEGRAMGEQLLPGKKFWETDESSKDGPKGIFLGDQWRDSAWGTSDHSVSQPIMVQRRPGQSFHVNSEVNSVLSPRSESGGLGVSMVEYVLSSSPGDSCLRKGGFGPRDADSDENDKGEKKNKGTFDGDKLGDLKEEGDVMDKTNGLPVQNGIDADVKDFSRTPGNCQNSANEVDLLGPNQNGSEGLAQLTSTNGAKPVEDFSNMESQSVPLDPMEHVGMEPLQFDYSGTQVPVDSAAATVGLFDYNSQQQLFQRPNALAVQQLTAAQQQQYALAAAHQPHIGLAPAAFVPNPYIISAAPPGTDPYTAGLAAAATLGPAVVPHQYYGVTPWGVYPASLFQQQAAAAAAATNSANQQTTPQAQQGQQQVLRGGASQRPLTPNQNQQGQQTDPLVAAAAVNSALAFGQGLAAGMPGYPVLAPAAYYDQTGALVVNAGARNGLGAPVRLVAPAPVIISSSAAQAAVAAAAASANGAAGGLAGTTNGPFRPLGTQQPQPQPQQQPNNNLASSSFYGNNSLNSNSQSSSLFSQGSAQPANTSLGFGSSSSLGATLGSALGGFGTAVANSNTGSGSRRDSLTGSSDLYKRTSSSLTPIGHSFYNGLSFSSSPGPVGMPLPSQGPGHSQTPPPSLSSHGSSSSLNLGGLTNGSGRYISAAPGAEAKYRSASSASSLFSPSSTLFSSSRLRYGMSDVMPSGRSRLLEDFRNNRYPNLQLREIAGHIMEFSQDQHGSRFIQLKLERATPAERQLVFNEILQAAYQLMVDVFGNYVIQKFFEFGSLEQKLALAERIRGHVLSLALQMYGCRVIQKALEFIPSDQQNEMVRELDGHVLKCVKDQNGNHVVQKCIECVQPQSLQFIIDAFKGQVFALSTHPYGCRVIQRILEHCLPDQTLPILEELHQHTEQLVQDQYGNYVIQHVLEHGRPEDKSKIVAEIRGNVLVLSQHKFASNVVEKCVTHASRTERAVLIDEVCTMNDGPHSALYTMMKDQYANYVVQKMIDVAEPGQRKIVMHKIRPHIATLRKYTYGKHILAKLEKYYMKNGVDLGPICGPPNGII

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service