Skip to Content
Merck
All Photos(2)

Key Documents

HPA006539

Sigma-Aldrich

Anti-SLC2A3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GLUT3, Anti-GTR3, Anti-Glucose transporter type 3, brain, Anti-Solute carrier family 2, facilitated glucose transporter member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

KVPETRGRTFEDITRAFEGQAHGADRSGKDGVMEMNSIEPAKETTT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC2A3(6515)

Immunogen

Solute carrier family 2, facilitated glucose transporter member 3 recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

SLC2A3 (solute carrier family 2) encodes a high-affinity glucose transporter protein. It is widely expressed in various tissues specifically chondrocytes. It is involved in the immune response and chondrocyte metabolism. During embryonic development, it helps to transport glucose from maternal blood across the placental trophoblastic tissue for the fetal growth. In rheumatoid arthritis (RA), it plays a vital role in the immune response and chondrocyte function. In addition to RA, it is also associated with various diseases, including dyslexia, Alzheimer′s disease, schizophrenia, and Huntingtons disease. It has also been reported that GLUT3 gene may have clinical importance in the myelomeningocele (MM) disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70035

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brendan D Connealy et al.
American journal of obstetrics and gynecology, 211(3), 305-305 (2014-05-13)
Our objectives were to examine the extent of described sequence variation in the glucose transporter 3 (GLUT3) gene in children with myelomeningocele (MM), identify novel variations in the GLUT3 gene in these children, and determine whether these variations may confer
Shaojun Yun et al.
Toxicology letters, 310, 23-30 (2019-04-14)
The aim of this study was to determine whether Pb affects glucose metabolism in the hippocampus of rats. Male Sprague-Dawley rats aged 21 days were orally administered a 0.1%, 0.2%, or 0.3% lead acetate solution in deionized water for 65
Taichi Noda et al.
Biology of reproduction, 100(4), 1035-1045 (2018-11-20)
Seminal vesicle secretions (SVSs), together with spermatozoa, are ejaculated into the female reproductive tract. SVS7, also known as PATE4, is one of the major SVS proteins found in the seminal vesicle, copulatory plug, and uterine fluid after copulation. Here, we
Colin D Veal et al.
Human mutation, 35(2), 248-256 (2013-11-02)
We describe a copy-number variant (CNV) for which deletion alleles confer a protective affect against rheumatoid arthritis (RA). This CNV reflects net unit deletions and expansions to a normal two-unit tandem duplication located on human chr12p13.31, a region with conserved
Kelin Gonçalves de Oliveira et al.
Acta neuropathologica communications, 12(1), 35-35 (2024-02-28)
Immunotherapies with antibody-drug-conjugates (ADC) and CAR-T cells, targeted at tumor surface antigens (surfaceome), currently revolutionize clinical oncology. However, target identification warrants a better understanding of the surfaceome and how it is modulated by the tumor microenvironment. Here, we decode the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service