Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300006

Sigma-Aldrich

Anti-PRDM16 (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-PFM13, MEL1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (i)
IHC
clone:
polyclonal
species reactivity:
human
citations:
6
technique(s):
immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRDM16(63976)

General description

PR/SET domain 16 (PRDM16) is encoded by the gene mapped to human chromosome 1p36.32. PRDM16 is a 140kDa zinc-finger protein characterized with a PR (PRD1-BF1-RIZ1 homologous)-domain. It is highly expressed in brown fat cells compared to white fat cells.
The reciprocal translocation t(1;3)(p36;q21) occurs in a subset of myelodysplastic syndrome (MDS) and acute myeloid leukemia (AML). This gene is located near the 1p36.3 breakpoint and has been shown to be specifically expressed in the t(1:3)(p36,q21)-positive MDS/AML. The protein encoded by this gene is a zinc finger transcription factor and contains an N-terminal PR domain. The translocation results in the overexpression of a truncated version of this protein that lacks the PR domain, which may play an important role in the pathogenesis of MDS and AML. Alternatively spliced transcript variants encoding distinct isoforms have been reported. Purified recombinant GST fusion protein encoding N-terminal aa 13-316 of human PRDM16.

Immunogen

PRDM16 (NP_071397, 1-350)
Purified recombinant GST fusion protein encoding N-terminal aa 13-316 of human PRDM16.

Application

Anti-PRDM16 (N-term) antibody produced in rabbit has been used in western blotting..

Biochem/physiol Actions

PR/SET domain 16 (PRDM16) is a potential transcriptional regulator. It regulates a bidirectional cell fate switch between skeletal myoblasts and brown fat cells. The encoded protein plays an essential role in promoting adipogenesis by co-activating transcriptional function of PPARγ. PRDM16 is required for cardiac development. Thus, loss of PRDM16 gene expression leads to the development of cardiomyopathy of 1p36 deletion syndrome. Variation in the gene expression is associated with the development of migraine.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transcriptional Control of Brown Fat Determination by PRDM16
Seale P, et al.
Cell Metabolism, 6(1), 38-54 (2007)
Loss of PRDM16 Is Unlikely to Cause Cardiomyopathy in 1p36 Deletion Syndrome
de Leeuw N, et al.
American Journal of Human Genetics, 94(1), 153-154 (2014)
Cdkn1c boosts the development of brown adipose tissue in a murine model of Silver Russell syndrome.
Van De Pette M, et al.
PLoS Genetics, 12(3), e1005916-e1005916 (2016)
PRDM16 Controls a Brown Fat/Skeletal Muscle Switch
Seale P, et al.
Nature, 454(7207), 961-961 (2008)
Genome-wide Association Study Reveals Three Susceptibility Loci for Common Migraine in the General Population
Chasman D I, et al.
Nature Genetics, 43(7), 695-695 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service