Skip to Content
Merck
All Photos(5)

Key Documents

HPA014405

Sigma-Aldrich

Anti-UGT8 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-2-hydroxyacylsphingosine 1-beta-galactosyltransferase precursor, Anti-Ceramide UDP-galactosyltransferase, Anti-Cerebroside synthase, Anti-UDP-galactose-ceramide galactosyltransferase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

SALHERGHHTVFLLSEGRDIAPSNHYSLQRYPGIFNSTTSDAFLQSKMRNIFSGRLTAIELFDILDHYTKNCDLMVGNHALIQGLKKEKFDLLLVDPNDMCGFVIAHLLGVKYAVFSTGLWYPAEVGAPA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UGT8(7368)

General description

The gene UGT8 (UDP glycosyltransferase 8) encodes a biosynthetic member of the UDP-glycosyltransferase family of proteins. It is abundantly expressed in the kidney and gastrointestinal tract. The human UGT superfamily consists of four families of enzymes that participate in the addition of sugar residues to small lipophilic chemicals. The gene is mapped to human chromosome 4q26.

Immunogen

2-hydroxyacylsphingosine 1-beta-galactosyltransferase precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene UGT8 (UDP glycosyltransferase 8) encodes an enzyme that catalyzes the conversion of UDP galactose to galactosidate ceramide, an important step in the synthesis of brain sphingolipids. It also galactosidates bile acids and drug-like bile acid analogs. In vitro studies show that UGT8 conjugates bile acids approximately 60 times more efficiently than ceramide. It serves as a potential prognostic marker for lung metastases in breast cancer and tumor aggressiveness.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72535

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Lionetti et al.
Genes, chromosomes & cancer, 48(6), 521-531 (2009-03-24)
It is thought that altered microRNA (miRNA) expression due to various mechanisms plays a critical role in the pathogenesis of most human cancers. Notably, about half of the known miRNAs are intragenic and frequently coexpressed with their host genes. To
Tomasz B Owczarek et al.
PloS one, 8(12), e84191-e84191 (2014-01-07)
It was recently proposed that UDP-galactose:ceramide galactosyltransferase (UGT8), enzyme responsible for synthesis of galactosylceramide (GalCer), is a significant index of tumor aggressiveness and a potential marker for the prognostic evaluation of lung metastases in breast cancer. To further reveal the
Robyn Meech et al.
Molecular pharmacology, 87(3), 442-450 (2014-12-19)
The human UDP glycosyltransferase (UGT) superfamily comprises four families of enzymes that catalyze the addition of sugar residues to small lipophilic chemicals. The UGT1 and UGT2 enzymes use UDP-glucuronic acid, and UGT3 enzymes use UDP-N-acetylglucosamine, UDP-glucose, and UDP-xylose to conjugate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service