Skip to Content
Merck
All Photos(1)

Key Documents

S8951

Sigma-Aldrich

SR11237

≥98% (HPLC)

Synonym(s):

4-[2-(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)-1,3-dioxolan-2-yl]-benzoic acid, BMS 188649, BMS649

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H28O4
CAS Number:
Molecular Weight:
380.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >28 mg/mL (warmed)

storage temp.

2-8°C

SMILES string

CC1(C)CCC(C)(C)c2cc(ccc12)C3(OCCO3)c4ccc(cc4)C(O)=O

InChI

1S/C24H28O4/c1-22(2)11-12-23(3,4)20-15-18(9-10-19(20)22)24(27-13-14-28-24)17-7-5-16(6-8-17)21(25)26/h5-10,15H,11-14H2,1-4H3,(H,25,26)

InChI key

ZZUKALQMHNSWTK-UHFFFAOYSA-N

Application

SR11237 may be used in RXR-mediated cell signaling studies.

Biochem/physiol Actions

SR11237 and other ligands of retinoid X receptor (RXR) activate various nuclear receptors during development process. This produces malformations in Xenopus embryos, involving the anterior-posterior axis.1
SR11237 is a selective pan retinoid X receptor (RXR) agonist with no retinoid A receptor (RAR) activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection of a receptor-ligand non-covalent complex using a triple quadrupole mass spectrometer.
Johan Lengqvist et al.
Rapid communications in mass spectrometry : RCM, 16(20), 2003-2006 (2002-10-04)
W Peter Lippert et al.
ChemMedChem, 4(7), 1143-1152 (2009-06-06)
C/Si switch: Twofold sila-substitution (C/Si exchange) in the RXR-selective retinoids 4 a (SR11237) and 5 a leads to 4 b (disila-SR11237) and 5 b, respectively. Chemistry and biology of the C/Si pairs are reported.SR11237 (BMS649, 4 a) is a pan-RXR-selective
Guo-quan Chen et al.
International journal of cancer, 99(2), 171-178 (2002-04-30)
Epidemiological and animal studies have demonstrated that vitamin A and its natural and synthetic derivatives, retinoids, are effective agents in preventing the development of tobacco-associated cancers. Unfortunately, clinical trials of retinoids on cigarette smokers have shown lack of efficacy in
G J Gendimenico et al.
The Journal of investigative dermatology, 102(5), 676-680 (1994-05-01)
We evaluated SR11237, a retinoid X receptor (RXR)-specific compound, for its pharmacologic effects on cell differentiation in F9 embryonal carcinoma cells and rhino mouse epidermis. SR11237 can cause RXR/RXR homodimers to form and transactivate a reporter gene containing a RXR-response
G Benoit et al.
The EMBO journal, 18(24), 7011-7018 (1999-12-22)
Although retinoic acid receptor alpha (RARalpha) agonists induce the maturation of t(15;17) acute promyelocytic leukemia (APL) cells, drug treatment also selects leukemic blasts expressing PML-RARalpha fusion proteins with mutated ligand-binding domains that no longer respond to all-trans retinoic acid (ATRA).

Articles

We offer many products related to non-steroid nuclear receptors for your research needs.

We offer many products related to non-steroid nuclear receptors for your research needs.

We offer many products related to non-steroid nuclear receptors for your research needs.

We offer many products related to non-steroid nuclear receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service