S7942
Sirtinol
≥95% (HPLC)
Synonym(s):
2-[(2-Hydroxynaphthalen-1-ylmethylene)amino]-N-(1-phenethyl)benzamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C26H22N2O2
CAS Number:
Molecular Weight:
394.47
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.77
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
solid
solubility
DMSO: soluble
storage temp.
−20°C
SMILES string
CC(NC(=O)c1ccccc1\N=C\c2c(O)ccc3ccccc23)c4ccccc4
InChI
1S/C26H22N2O2/c1-18(19-9-3-2-4-10-19)28-26(30)22-13-7-8-14-24(22)27-17-23-21-12-6-5-11-20(21)15-16-25(23)29/h2-18,29H,1H3,(H,28,30)/b27-17+
InChI key
UXJFDYIHRJGPFS-WPWMEQJKSA-N
Gene Information
human ... SIRT1(23411) , SIRT2(22933)
Application
HUVEC were treated with sirtinol to study the effect on induction of transcription factor Krüppel-like factor 2.4 Sirtinol was used to treat human embryonic kidney cells to study the effect on the expression on Werner syndrome protein.5
Biochem/physiol Actions
Sirtinol inhibits yeast Sir2p transcriptional silencing activity in vivo, yeast Sir2p and human SIRT2 deacetylase activity in vitro. It inhibits the physiological regulators of platelet aggregation such as thrombin and collagen, attenuates intracellular Ca2+ release and formation thromboxane B2. It may increase levels of cAMP by inhibition of cAMP phosphodiesterase and inhibit the aggregation of platelets.2 Sirtinol reduces inflammatory responses of human dermal microvascular endothelial cells to TNF-α and IL-1β.3
Sirtinol inhibits yeast Sir2p transcriptional silencing activity in vivo, yeast Sir2p, and human SIRT2 deacetylase activity in vitro.
Features and Benefits
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
H Mutoh et al.
The American journal of physiology, 261(1 Pt 1), G65-G70 (1991-07-01)
We examined the role of reduced glutathione as a defense mechanism against acid-induced gastric mucosal cell damage in vitro. Cellular stores of reduced glutathione were depleted by reaction with diethyl maleate (DEM) or 1-chloro-2,4-dinitrobenzene (CDNB) and increased by reaction with
Angela Orecchia et al.
PloS one, 6(9), e24307-e24307 (2011-09-21)
Histone deacetylases (HDAC) are key enzymes in the epigenetic control of gene expression. Recently, inhibitors of class I and class II HDAC have been successfully employed for the treatment of different inflammatory diseases such as rheumatoid arthritis, colitis, airway inflammation
Na-Young Song et al.
Biochemical pharmacology, 153, 282-291 (2018-02-11)
Leptin, a representative adipokine secreted from the white adipose tissue, is considered as a potential linker between obesity and cancer. SIRT1 is an NAD+-dependent histone/protein deacetylase speculated to function as an oncogene. In the present study, we found that leptin
Dong Ryeol Ryu et al.
Aging cell, 18(2), e12904-e12904 (2019-01-08)
Although it is known that the expression and activity of sirtuin 1 (Sirt1) decrease in the aged kidney, the role of interaction between Sirt1 and hypoxia-inducible factor (HIF)-1α is largely unknown. In this study, we investigated whether HIF-1α could be
Jun Feng et al.
European journal of pharmacology, 791, 632-639 (2016-10-30)
Tanshinone IIA (Tan) exerts potential protective effects against cardiovascular diseases. Oxidative stress and inflammation are involved in cardiac hypertrophy. Activation of silent information regulator 1 (SIRT1) signaling has been suggested to attenuate cardiac hypertrophy. This study aims to evaluate the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service