S7023
Sera human
frozen liquid
Synonym(s):
Human Blood Serum, Human Serum
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
Quality Level
form
frozen liquid
origin
USA origin
shipped in
dry ice
storage temp.
−20°C
General description
The serum is the fluid from plasma devoid of fibrinogen, prothrombin, and other clotting factors. It contains various plasma proteins, amino acids, polypeptides, growth factors, fat, carbohydrates, nucleic acid derivatives, hormones, inorganic minerals, and vitamins.
Application
Sera human has been used:
- to prepare the antigen standards for the validation assay of multiple-biomarker detection
- as a quality check (QC) sample to optimize the experimental workflow
- as a control in immunoproteomics assay
Biochem/physiol Actions
Human serum is used as a cell culture supplement. It maintains physiological balance by stimulating or inhibiting cell growth. It is known to have an inhibitory action on endogenous and exogenous enzymes.
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Microbiology spectrum, 9(1), e0102321-e0102321 (2021-08-26)
The production of capsular polysaccharides by Klebsiella pneumoniae protects the bacterial cell from harmful environmental factors such as antimicrobial compounds and infection by bacteriophages (phages). To bypass this protective barrier, some phages encode polysaccharide-degrading enzymes referred to as depolymerases to
Infection and immunity, 88(8) (2020-06-10)
The serum complement system is a first line of defense against bacterial invaders. Resistance to killing by serum enhances the capacity of Klebsiella pneumoniae to cause infection, but it is an incompletely understood virulence trait. Identifying and characterizing the factors
Nature protocols, 1(2), 880-891 (2007-04-05)
Blood is a convenient source of biomarkers. Readily obtainable, it immerses most tissues in the body and is therefore likely to contain cell-derived proteins and peptides that may provide information about various biological processes. Serum proteome and peptidome profiling--using mass
The Journal of molecular diagnostics : JMD, 19(2), 303-312 (2017-01-04)
Extraction of viral RNA and the storage of sample material are extremely important factors in the detection and whole genome sequencing (WGS) of viral pathogens. Although PCR-based detection methods focus on small amplicons, viral WGS applications require RNA of high
Vaccines, 8(3) (2020-09-10)
Infections by the Burkholderia cepacia complex (Bcc) remain seriously life threatening to cystic fibrosis (CF) patients, and no effective eradication is available. A vaccine to protect patients against Bcc infections is a highly attractive therapeutic option, but none is available.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service