Skip to Content
Merck
All Photos(1)

Key Documents

H6914

Sigma-Aldrich

Human Serum

(from male AB clotted whole blood), USA origin, sterile-filtered

Synonym(s):

human sera, human serum, off the clot, otc

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

biological source

human

Quality Level

sterility

sterile-filtered

composition

Hemoglobin, ≤25 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable
cell culture | stem cell: suitable

impurities

≤10 EU/mL Endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Human AB serum is used in tissue engineering, transplantation and cell therapy applications for the expansion of mesenchymal stem cells (MSC) from adipose tissue or mesenchymal stromal cells from human bone marrow; for standardized limbal epithelial stem cell graft generation and transplantation; for ex vivo expansion of NK cells from peripheral blood in Hematopoeitic Stem Cell Expansion Medium; and for upgrading pretransplant human islet culture technology.

Packaging

Packaging may vary for different batches due to short supply of the bottles used to pack this product. The presence or lack of shrink wrap around the cap of the bottle doesn’t impact product integrity/sterility of the product.

Warning

Each donor is tested for and found non-reactive for Hepatitis B & C and non-reactive for Human Immunodeficiency Virus (HIV) antibody by ELISA. Nevertheless, products of human origin should be considered potentially infectious and handled accordingly.

Analysis Note

Endotoxin tested

Other Notes

It is not unusual for our human serum products to be cloudy. This product is from pooled serum and cloudiness indicates that lipids are present in the serum. No clear, lipid-free human serum products are available.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alejandro Bustamante et al.
Stroke, 48(9), 2419-2425 (2017-07-19)
Stroke diagnosis could be challenging in the acute phase. We aimed to develop a blood-based diagnostic tool to differentiate between real strokes and stroke mimics and between ischemic and hemorrhagic strokes in the hyperacute phase. The Stroke-Chip was a prospective
Kwang Hoe Kim et al.
Analytica chimica acta, 882, 38-48 (2015-06-05)
Multiple reaction monitoring (MRM) is commonly used for the quantitative analysis of proteins during mass pectrometry (MS), and has excellent specificity and sensitivity for an analyte in a complex sample. In this study, a pseudo-MRM method for the quantitative analysis
Ankit Sinha et al.
Cell systems, 8(4), 345-351 (2019-04-15)
High-grade serous ovarian carcinoma (HGSC) is the most common and lethal subtype of gynecologic malignancy in women. The current standard of treatment combines cytoreductive surgery and chemotherapy. Despite the efficacy of initial treatment, most patients develop cancer recurrence, and 70%
Laura Abraira et al.
Epilepsia, 61(10), 2244-2253 (2020-08-29)
Blood biomarkers have not been widely investigated in poststroke epilepsy. In this study, we aimed to describe clinical factors and biomarkers present during acute stroke and analyze their association with the development of epilepsy at long term. A panel of
Hien T Ngoc Le et al.
Micromachines, 11(9) (2020-08-23)
Amyloid beta (aβ) 1-42, a peptide that is 1-42 amino acids long, is a major component of senile plaques in the brains of patients with Alzheimer's disease. Aβ detection has become an essential antecedence to predict the declining mental abilities

Articles

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service