Skip to Content
Merck
All Photos(4)

Key Documents

S5011

Sigma-Aldrich

Sodium phosphate monobasic

BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)

Synonym(s):

Monosodium dihydrogen orthophosphate, Monosodium phosphate, Sodium dihydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH2PO4
CAS Number:
Molecular Weight:
119.98
EC Number:
MDL number:
UNSPSC Code:
12352207
eCl@ss:
38070208
PubChem Substance ID:
NACRES:
NA.75

grade

BioPerformance Certified

Quality Level

Assay

≥99.0% (titration)

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable
cell culture | plant: suitable

pH

4.0-4.5 (25 °C, 50 g/L in water)

SMILES string

[Na+].OP(O)([O-])=O

InChI

1S/Na.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

AJPJDKMHJJGVTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Monosodium phosphate (NaH2PO4), a phosphatizing agent is used on steel surfaces. It is soluble in water.

Application

Sodium phosphate monobasic has been used:
  • in insect cell culture to adjust pH
  • as a component in culture medium for pig oocyte maturation and fertilization
  • as a resealing solution for electroporation

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PHOSPHORUS| Properties and Determination
Garg AN
Encyclopedia of food sciences and nutrition (2003)
Characterization of carrier erythrocytes for biosensing applications
Lopez SCB and Meissner KE
Journal of Biomedical Optics, 22(9), 091510-091510 (2017)
Abundant DNA 6mA methylation during early embryogenesis of zebrafish and pig.
Liu J et al.
Nature Communications, 7, 13052-13052 (2016)
Jianzhao Liu et al.
Nature communications, 7, 13052-13052 (2016-10-08)
DNA N6-methyldeoxyadenosine (6mA) is a well-known prokaryotic DNA modification that has been shown to exist and play epigenetic roles in eukaryotic DNA. Here we report that 6mA accumulates up to ∼0.1-0.2% of total deoxyadenosine during early embryogenesis of vertebrates, but
Michael Zabolocki et al.
Nature communications, 11(1), 5550-5550 (2020-11-05)
The capabilities of imaging technologies, fluorescent sensors, and optogenetics tools for cell biology are advancing. In parallel, cellular reprogramming and organoid engineering are expanding the use of human neuronal models in vitro. This creates an increasing need for tissue culture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service