Skip to Content
Merck
All Photos(4)

Key Documents

S3139

Sigma-Aldrich

Sodium phosphate monobasic

BioReagent, for molecular biology, anhydrous, ≥98%

Synonym(s):

Monosodium dihydrogen orthophosphate, Monosodium phosphate, Sodium dihydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH2PO4
CAS Number:
Molecular Weight:
119.98
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
38070208
PubChem Substance ID:
NACRES:
NA.52

grade

anhydrous
for molecular biology

Quality Level

product line

BioReagent

Assay

≥98%

form

solid

technique(s)

activity assay: suitable

pH

4.0-4.5 (25 °C, 50 g/L in water)

solubility

H2O: complete 100 mg/mL, clear, colorless

anion traces

chloride (Cl-): ≤40 ppm

cation traces

Fe: ≤30 ppm

application(s)

diagnostic assay manufacturing

foreign activity

DNase, RNase and protease, none detected

storage temp.

room temp

SMILES string

[Na+].OP(O)([O-])=O

InChI

1S/Na.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

AJPJDKMHJJGVTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium phosphate is a salt of sodium hydroxide and phosphoric acid. It is widely used in buffers for molecular biology applications.

Application

Sodium phosphate monobasic is widely used in buffers for molecular biology applications. It is an important ingredient in cholesterol assay reaction buffer and miRNA microarray fabrication.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang-Gong Liu et al.
Methods (San Diego, Calif.), 44(1), 22-30 (2007-12-26)
After 12 years from its first application, microarray technology has become the reference technique to monitor gene expression of thousands of genes in the same experiment. In the past few years an increasing amount of evidence showed the importance of
Andrew D Pucker et al.
Investigative ophthalmology & visual science, 51(6), 2949-2954 (2010-01-22)
The purpose of this work was to develop an enzymatic method of quantification of cholesterol and cholesterol esters derived from contact lenses, both in vitro and ex vivo. Lotrafilcon B (O2 Optix; CIBA Vision, Inc., Duluth, GA) and galyfilcon A
Antje Dittmann et al.
Molecular systems biology, 15(8), e8849-e8849 (2019-08-30)
Obesity-associated type 2 diabetes and accompanying diseases have developed into a leading human health risk across industrialized and developing countries. The complex molecular underpinnings of how lipid overload and lipid metabolites lead to the deregulation of metabolic processes are incompletely
Menglin Liu et al.
Nature communications, 10(1), 2126-2126 (2019-05-11)
Repair of the endothelial cell barrier after inflammatory injury is essential for tissue fluid homeostasis and normalizing leukocyte transmigration. However, the mechanisms of endothelial regeneration remain poorly understood. Here we show that the endothelial and hematopoietic developmental transcription factor Sox17
Louise Thines et al.
Bio-protocol, 10(5), e3542-e3542 (2021-03-05)
Superoxide dismutases (SODs) act as a primary defence against reactive oxygen species (ROS) by converting superoxide anion radicals (O2 -) into molecular oxygen (O2) and hydrogen peroxide (H2O2). Members of this enzyme family include CuZnSODs, MnSODs, FeSODs, and NiSODs, depending

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service