S4651
Silane-Prep Slides
glass slides coated with silane (aminoalkylsilane).
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41122601
NACRES:
NA.47
Recommended Products
biological source
synthetic
application(s)
hematology
histology
storage temp.
room temp
General description
Silane-Prep Slides are microscope slides coated with silane (aminoalkysilane) to improve the adhesion of tissues to the slide for immunohistochemistry. The slide includes a "frosted end" for slide/patient identification. It is not tested for use in microarray assays.
Application
Silane-Prep Slides have been used in:
- Immunofluorescence microscopy of DNA fibers.
- Immunohistochemistry.
- The determination of hybridization stringency.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yuan-Ling Yu et al.
Insect science, 28(2), 315-329 (2020-02-29)
Rice stripe virus (RSV) is the causative agent of rice stripe disease and is completely dependent on insect vectors for its plant-to-plant transmission. Laodelphax striatellus is the major insect vector for RSV. In this study, we explored the interactions between
Prospects and Applications for Plant Associated Microbes. A Laboratory Manual, Part A: Bacteria (2008)
Skeletal muscle atrophy increases cell proliferation in mice gastrocnemius during the first week of hindlimb suspension
Ferreira R, et al.
European Journal of Applied Physiology, 97(3), 340-346 (2006)
Jorge I Alvarez et al.
PLoS neglected tropical diseases, 2(4), e218-e218 (2008-04-10)
Neurocysticercosis (NCC) is an infection of the central nervous system (CNS) by the metacestode of the helminth Taenia solium. The severity of the symptoms is associated with the intensity of the immune response. First, there is a long asymptomatic period
Rob P A Janssen et al.
Knee surgery, sports traumatology, arthroscopy : official journal of the ESSKA, 19(8), 1299-1306 (2011-02-05)
Histological analysis of the remodelling process of human hamstring tendon (HT) grafts after standardized anterior cruciate ligament reconstruction (ACLR) with an accelerated rehabilitation protocol. Sixty-seven patients underwent retrieval of mid-substance biopsies after clinically successful hamstring autograft ACLR. Samples were allocated
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service