Skip to Content
Merck
All Photos(3)

Key Documents

S4250

Sigma-Aldrich

D-Serine

≥98% (TLC), suitable for ligand binding assays

Synonym(s):

β-Hydroxyalanine, (R)-2-Amino-3-hydroxypropionic acid, Ser

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO3
CAS Number:
Molecular Weight:
105.09
Beilstein:
1721403
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

product name

D-Serine, ≥98% (TLC)

Quality Level

Assay

≥98% (TLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white to off-white

application(s)

cell analysis

SMILES string

N[C@H](CO)C(O)=O

InChI

1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m1/s1

InChI key

MTCFGRXMJLQNBG-UWTATZPHSA-N

Gene Information

human ... GRIN1(2902)

Looking for similar products? Visit Product Comparison Guide

General description

D-serine is a dextro amino acid present in neurons, astrocytes, and microglia. It is synthesized from L-serine by the enzyme serine racemase. Racemase also catalyzes the breakdown of D-serine into ammonia and pyruvate. It is oxidized to hydroxypyruvate by D-amino acid oxidase.
D-serine is an unusual amino acid expressed in the mammalian brain.

Application

D-serine has been used as a substrate in D-amino acid oxidase (DAO) activity in human 1321N1 astrocytoma cells. It has also been used in intracerebroventricular administration in rat for the induction of antinociceptive effect.
D-serine has been used to prevent glycine-dependent desensitization of N-methyl D-aspartate receptor (NMDAR) and to study its effects on NMDARs to correct behavioral abnormalities in rats after partial sciatic nerve ligation (PSNL).

Biochem/physiol Actions

D-serine is essential for the normal development of dendrites, neuroblast migration and may have therapeutic potential for treating schizophrenia and depression states. The levels of D-serine is elevated in traumatic brain injury (TBI).
D-serine is an agonist and glycine mimic which is active at the strychnine-insensitive glycine binding site associated with the N-methyl-D-aspartate (NMDA) receptor as well as the inhibitory post-synaptic glycine receptor. Along with glutamate, it has a role in various physiological processes including synaptic plasticity and receptor transmission. Dysregulation of D-serine signaling has been linked with neurodegenerative diseases and disorders.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Impact of Aging in Microglia-Mediated D-Serine Balance in the CNS
Beltran-Castillo S, et al.
Mediators of Inflammation, 2018 (2018)
D-amino acids in the brain: D-serine in neurotransmission and neurodegeneration.
Wolosker H
FEBS Journal, 275(14), 3514-3526 (2008)
Enhanced astrocytic d-serine underlies synaptic damage after traumatic brain injury
Perez EJ, et al.
The Journal of Clinical Investigation, 127(8), 3114-3125 (2017)
D-serine as a gliotransmitter and its roles in brain development and disease
Van Horn MR, et al.
Frontiers in Cellular Neuroscience, 7, 39-39 (2013)
Antinociceptive effect of intracerebroventricular administration of d-serine on formalin-induced pain
Ito M, et al.
Journal of Anesthesia, 28(2), 228-234 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service