Skip to Content
Merck
All Photos(4)

Key Documents

M3262

Sigma-Aldrich

N-Methyl-D-aspartic acid

≥98% (TLC), suitable for cell culture

Synonym(s):

(R)-2-(Methylamino)succinic acid, NMDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO4
CAS Number:
Molecular Weight:
147.13
Beilstein:
1724431
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.32

product name

N-Methyl-D-aspartic acid, ≥98% (TLC), solid

Quality Level

Assay

≥98% (TLC)

form

solid

technique(s)

cell culture | mammalian: suitable

color

white

mp

189-190 °C

application(s)

cell analysis

SMILES string

CN[C@H](CC(O)=O)C(O)=O

InChI

1S/C5H9NO4/c1-6-3(5(9)10)2-4(7)8/h3,6H,2H2,1H3,(H,7,8)(H,9,10)/t3-/m1/s1

InChI key

HOKKHZGPKSLGJE-GSVOUGTGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Methyl-D-aspartic acid (NMDA) is an endogenously generated molecule in rat nervous system and endocrine glands. NMDA is present at low level (nmol/g) in the adenohypophysis, hypothalamus, brain, and testis. NMDA is derived from D-Asp by an S-adenosylmethionine-dependent enzyme also referred to as NMDA synthase.

Application

N-Methyl-D-aspartic acid has been used to induce N-methyl-d-aspartic acid (NMDA) toxicity, cultures of retinal cells were supplemented with NMDA.

Biochem/physiol Actions

Excitotoxic amino acid. Prototypic agonist at the NMDA-type glutamate receptor that regulates ion channels; important in long-term potentiation, ischemia, and epilepsy.
N-Methyl-D-aspartic acid (NMDA) plays a crucial role in release of luteinizing hormone and PRL (prolactin) in the pituitary gland and GnRH (gonadotropin releasing hormone) in the hypothalamus. It acts as a specific agonist for NMDA type glutamate receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calpain- and caspase-mediated aII-spectrin
and tau proteolysis in rat cerebrocortical
neuronal cultures after ecstasy or
methamphetamine exposure.
Warren MW
The International Journal of Neuropsychopharmacology, 9, 1-11 (2006)
Guendalina Olivero et al.
Molecular neurobiology, 56(9), 6142-6155 (2019-02-09)
Mouse hippocampal glutamatergic nerve endings express presynaptic release-regulating NMDA autoreceptors (NMDARs). The presence of GluN1, GluN2A, GluN2B, and GluN3A subunits in hippocampal vesicular glutamate transporter type 1-positive synaptosomes was confirmed with confocal microscopy. GluN2C, GluN2D, and GluN3B immunopositivity was scarcely
Occurrence of D-aspartic acid and N-methyl-D-aspartic acid in rat neuroendocrine tissues and their role in the modulation of luteinizing hormone and growth hormone release.
D'Aniello A
Faseb Journal, 14(5), 699-714 (2000)
Juan Ding et al.
Neural regeneration research, 14(12), 2112-2117 (2019-08-10)
N-methyl-D-aspartate receptor hypofunction is the basis of pathophysiology in schizophrenia. Blocking the N-methyl-D-aspartate receptor impairs learning and memory abilities and induces pathological changes in the brain. Previous studies have paid little attention to the role of the N-methyl-D-aspartate receptor subunit
N-methyl-D-aspartic acid (NMDA) in the nervous system of the amphioxus Branchiostoma lanceolatum.
D'Aniello S
BMC Neuroscience, 8:109 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service