Skip to Content
Merck
All Photos(3)

Key Documents

S3132

Sigma-Aldrich

Seleno-L-methionine

≥98% (TLC), powder, IgE expression inhibitor

Synonym(s):

(S)-2-Amino-4-(methylseleno)butyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SeCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
196.11
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

product name

Seleno-L-methionine, ≥98% (TLC), powder

Quality Level

Assay

≥98% (TLC)

form

powder

color

white to off-white

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

C[Se]CC[C@H](N)C(O)=O

InChI

1S/C5H11NO2Se/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1

InChI key

RJFAYQIBOAGBLC-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Seleno-L-Methionine (SeMet), an organic form of selenium is found in Cupriavidus metallidurans.

Application

Seleno-L-methionine has been used:
  • in toxicity studies in Japanese Medaka embryos
  • to study its effects on selenium status indicators in pregnant long-tailed macaques (Macaca Fascicularis)
  • to label proteins for single wavelength anomalous dispersion (SAD) phasing

Seleno-L-methionine has been used:
  • as a nonchemical stressor to treat Japanese Medaka fish embryos
  • to label proteins for Single wavelength Anomalous Dispersion (SAD) phasing
  • to study the distribution and inhibition effects of seleno-L-methionine on 4T1 mouse mammary carcinoma

Biochem/physiol Actions

Seleno-L-Methionine (SeMet) is capable of repressing atopic dermatitis (AD)-like skin lesions. It can also prevent the expression of total immunoglobulin E (IgE) and interleukin 4 (IL-4). It can cause ecotoxicity as it is absorbed by fish through diet.
Seleno-L-methionine displays antioxidant activity and has been shown to increase the activity of glutathione peroxidase in endothelial cells. Glutathione peroxidase protects cells from oxidative damage, such as DNA strand breaks, mutations and interference with protein tyrosine-based signaling and other protein functions due to formation of 3-nitrotyrosine, caused by excessive peroxynitrite. Seleno-L-methionine administration to cancer cell lines (MCF-7/S breast carcinoma, DU-145 prostate cancer cells and UACC-375 melanoma) results in apoptotic cell death and aberrant mitosis. These human tumor cell lines exhibited dose-dependent growth inhibition by selenomethionine in the micromolar range (45 to 130μM), while growth inhibition of normal fibroblasts required 1mM selenomethionine.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

250.3 - 346.3 °F

Flash Point(C)

121.3 - 174.6 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seleno-L-methionine is the predominant organic form of selenium in Cupriavidus metallidurans CH34 exposed to selenite or selenate
Avoscan L, et al.
Applied and Environmental Microbiology, 72(9), 6414-6416 (2006)
Arka Chakraborty et al.
Nucleic acids research, 45(2), 951-967 (2016-12-03)
The mitochondrial genome (mtDNA) is assembled into nucleo-protein structures termed nucleoids and maintained differently compared to nuclear DNA, the involved molecular basis remaining poorly understood. In yeast (Saccharomyces cerevisiae), mtDNA is a ∼80 kbp linear molecule and Abf2p, a double
H Sies et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 53(3-4), 228-232 (1998-06-10)
Cellular defense against excessive peroxynitrite generation is required to protect against DNA strand-breaks and mutations and against interference with protein tyrosine-based signaling and other protein functions due to formation of 3-nitrotyrosine. We recently demonstrated a role of selenium-containing enzymes catalyzing
A protocol to retain weakly interacting protein complexes for structural studies using an appropriate linker
Chichili VPR, et al.
Protocol Exchange (2013)
C Redman et al.
Cancer letters, 125(1-2), 103-110 (1998-05-05)
Selenium supplementation has been shown for many years to work as an anticarcinogenic agent both in epidemiology and in in vitro studies. Selenium supplementation has recently been shown to decrease total cancer incidence. However, the mechanism of action of selenium

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service