Skip to Content
Merck
All Photos(2)

Key Documents

545996

Sigma-Aldrich

Seleno-L-cystine

95%, for peptide synthesis

Synonym(s):

(R,R)-3,3′-Diseleno-bis(2-aminopropionic acid), L-Selenocystine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CO2HCH(NH2)CH2(Se)2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
334.09
Beilstein:
1969559
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

Seleno-L-cystine, 95%

Quality Level

Assay

95%

form

powder or crystals

optical activity

[α]20/D −28°, c = 1 in NaOH

reaction suitability

reaction type: solution phase peptide synthesis

mp

224.5-229.5 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

N[C@@H](C[Se][Se]C[C@H](N)C(O)=O)C(O)=O

InChI

1S/C6H12N2O4Se2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m0/s1

InChI key

JULROCUWKLNBSN-IMJSIDKUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Seleno-L-cystine can be used for the synthesis of:
  • Biologically active selenol compounds.[1]
  • Non-natural selenylated diamino acids.[2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Andreadou et al.
Journal of medicinal chemistry, 39(10), 2040-2046 (1996-05-10)
Eighteen Se-substituted selenocysteine derivatives were synthesized as potential kidney selective prodrugs which can be activated by renal cysteine conjugate beta-lyase to selenium-containing chemoprotectants or antitumor agents. Selenocysteine derivatives with aliphatic and benzylic Se-substituents were synthesized by reducing selenocystine to selenocysteine
Andrzej Gawor et al.
Animals : an open access journal from MDPI, 10(5) (2020-05-13)
Identification and quantification of the selenium species in biological tissues is imperative, considering the need to properly understand its metabolism and its importance in various field of sciences, especially nutrition science. Although a number of studies deals with the speciation
Synthesis of novel Se-substituted selenocysteine derivatives as potential kidney selective prodrugs of biologically active selenol compounds: evaluation of kinetics of ?-elimination reactions in rat renal cytosol
Andreadou I, et al.
Journal of Medicinal Chemistry, 39(10), 2040-2046 (1996)
Novel selenium-containing non-natural diamino acids
Caputo R, et al.
Tetrahedron Letters, 48(8), 1425-1427 (2007)
Erin E Battin et al.
Metallomics : integrated biometal science, 3(5), 503-512 (2011-02-03)
Copper and iron are two widely studied transition metals associated with hydroxyl radical (˙OH) generation, oxidative damage, and disease development. Because antioxidants ameliorate metal-mediated DNA damage, DNA gel electrophoresis assays were used to quantify the ability of ten selenium-containing compounds

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service