Skip to Content
Merck
All Photos(2)

Key Documents

S0697

Sigma-Aldrich

Anti-Sortilin antibody produced in rabbit

lyophilized powder, affinity isolated antibody

Synonym(s):

Anti-SORT1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry: suitable
western blot: 1:200 using rat brain lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SORT1(6272)
mouse ... Sort1(20661)
rat ... Sort1(83576)

Immunogen

peptide DKDTTRRIHVSTDQD(C) corresponding to residues 320-335 of human sortilin.

Target description

Sortilin is neurotensin receptor 3 (NTS3), one of the receptors for the peptide neurotransmitter neurotensin that exerts several biological functions ranging from the regulation of dopamine transmission and pain in the central nervous system to its functioning as a local hormone affecting the gastrointestinal tract. Sortilin expression was found to be essential for proNGF and proBDNF-induced neuronal cell death

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.025% sodium azide.

Reconstitution

Reconstitute the lyophilized vial with 50 μL or 200 μL deionized water, depending on package size. Further dilutions should be made using a carrier protein such as BSA (1-3%).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margalida Cifre et al.
Molecular neurodegeneration, 13(1), 14-14 (2018-03-24)
Metabolically-obese, normal-weight (MONW) individuals are not obese in terms of weight and height but have a number of obesity-related features (e.g. greater visceral adiposity, insulin resistance, and increased risk of cardiovascular disease). The MONW phenotype is related to the intake
Gregory T Jones et al.
Human molecular genetics, 22(14), 2941-2947 (2013-03-29)
Abdominal aortic aneurysm (AAA) is a common human disease with a high estimated heritability (0.7); however, only a small number of associated genetic loci have been reported to date. In contrast, over 100 loci have now been reproducibly associated with
Idy H T Ho et al.
Theranostics, 9(6), 1651-1665 (2019-05-01)
Rationale: Brain-derived neurotrophic factor (BDNF) is a key mediator in the development of chronic pain. Sortilin is known to interact with proBDNF and regulate its activity-dependent secretion in cortical neurons. In a rat model of inflammatory pain with intraplantar injection
Yi Xia et al.
International journal of molecular sciences, 14(7), 14085-14104 (2013-07-25)
Growing evidence has shown that proNGF-p75NTR-sortilin signaling might be a crucial factor in neurodegeneration, but it remains unclear if it may function in nigral neurons under aging and disease. The purpose of this study is to examine and quantify proNGF

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service