Skip to Content
Merck
All Photos(1)

Key Documents

RES3041T

SAFC

TMP disodium salt

Synonym(s):

Thymolphthalein monophosphate disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H29Na2O7P · xH2O
CAS Number:
Molecular Weight:
554.48 (anhydrous basis)
Beilstein:
7616883
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

form

powder

impurities

Free Thymolphthalein: tested

storage temp.

−20°C

SMILES string

[Na+].[Na+].[P](=O)([O-])([O-])Oc1c(cc(c(c1)C)C3(OC(=O)c4c3cccc4)c2c(cc(c(c2)C(C)C)O)C)C(C)C.O

InChI

1S/C28H31O7P.2Na.H2O/c1-15(2)20-13-23(17(5)11-25(20)29)28(22-10-8-7-9-19(22)27(30)34-28)24-14-21(16(3)4)26(12-18(24)6)35-36(31,32)33;;;/h7-16,29H,1-6H3,(H2,31,32,33);;;1H2/q;2*+1;/p-2

InChI key

VXAAAGIFNPRCKZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service