Skip to Content
Merck
All Photos(1)

Key Documents

71950

Sigma-Aldrich

Sodium selenite

anhydrous, ≥90.0% (RT)

Synonym(s):

Selenious acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SeO3
CAS Number:
Molecular Weight:
172.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥90.0% (RT)
form:
powder
Pricing and availability is not currently available.

Quality Level

Assay

≥90.0% (RT)

form

powder

loss

≤0.5% loss on drying, 200 °C

mp

>350 °C (lit.)

SMILES string

[Na+].[Na+].[O-][Se]([O-])=O

InChI

1S/2Na.H2O3Se/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

BVTBRVFYZUCAKH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium selenite can be used:
  • In dehydrogenation of benzoins to benzils and, flavanones to flavones under thermal and microwave conditions.[1]
  • As a catalyst along with cysteine or glutathione in the reductive debromination of vic-dibromides to alkenes.[2]
  • To prepare bis(methylmercuric) selenide (BMS) by reacting with methylmercuric chloride and reduced glutathione or cysteine.[3]

Sodium selenite is a common source of selenium in the synthesis of zinc selenide (ZnSe).[4] It is used as a key precursor in the synthesis of bismuth selenide nanocomposites,[5] nickel cobalt selenides,[6] selenium nanobelts[7] and t-selenium nanotubes.[8]

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sodium selenite-dimethylsulfoxide: a highly efficient reagent for dehydrogenation
Lamba M and Makrandi JK
J. Chem. Res. (M), 2008(4), 225-226 (2008)
Cellulose-directed growth of selenium nanobelts in solution.
Lu Q, et al.
Chemistry of Materials, 18(1) (2006)
Hydrothermal synthesis and high photocatalytic activity of 3D wurtzite ZnSe hierarchical nanostructures.
Cao F, et al.
The Journal of Physical Chemistry C, 112(44), 17095-17101 (2008)
Nucleation- dissolution- recrystallization: a new growth mechanism for t-selenium Nanotubes.
Xi G, et al.
Crystal Growth & Design, 6(2), 577-582 (2006)
Selenium-catalysed debromination of vic-dibromides to alkenes with cysteine or glutathione
Yanada, Kazuo and Yanada, Reiko and Meguri, Haruo
Journal of the Chemical Society. Chemical Communications, 2008(10), 730-732 (1990)

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service