Skip to Content
Merck
All Photos(1)

Key Documents

RAB0750

Sigma-Aldrich

Human PLAUR / Urokinase Plasminogen Activator Surface Receptor ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 15 pg/mL
standard curve range: 16.38-4000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PLAUR(5329)

General description

The antibody pair provided in this kit recognizes human Urokinase Plasminogen Activator Surface Receptor.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Plasminogen activator, urokinase receptor (PLAUR) is implicated in tumor invasion, angiogenesis and metastasis. It acts as a key regulator of cell-surface plasminogen activator. It is involved in various normal and pathological processes. Overexpression of the gene has been observed in various types of cancers, including head and neck squamous cell carcinoma (HNSCC). Thus, it is considered as an potent biomarker for targeted imaging or therapy. Elevated expression of PLAUR leads to the progression of kidney disease in children.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Huang et al.
Mediators of inflammation, 2020, 7850179-7850179 (2020-04-24)
The soluble urokinase plasminogen activator receptor (suPAR) is associated with kidney diseases and is used as a prognostic factor of renal function progression. The aim of this study was to explore whether circulating suPAR was associated with antineutrophil cytoplasmic autoantibody-
Ümmügülsüm Can et al.
Journal of investigative medicine : the official publication of the American Federation for Clinical Research, 62(7), 938-943 (2014-08-16)
Chronic obstructive pulmonary disease (COPD) results from an abnormal inflammatory response of the lungs to noxious particles or gases. Serum soluble urokinase-type plasminogen activator receptor (suPAR) is a glycoprotein secreted during infections and inflammation. The main goal of this study
Sven Wach et al.
Molecular therapy. Nucleic acids, 16, 272-283 (2019-04-02)
MIR143 is pathologically downregulated and may function as a tumor suppressor in prostate cancer. Likewise, the urokinase plasminogen activator receptor (UPAR) is overexpressed in prostate carcinoma, representing a negative prognostic marker and putative therapeutic target gene. In this paper, we
Assignment of the Urokinase-Type Plasminogen Activator Receptor Gene (PLAUR) to Chromosome 1 9q 13.1 -q 13.2
B?rglum A D, et al.
American Journal of Human Genetics, 50(3), 492-492 (1992)
uPAR-targeted optical near-infrared (NIR) fluorescence imaging and PET for image-guided surgery in head and neck cancer: proof-of-concept in orthotopic xenograft model
Christensen A, et al.
Oncotarget, 8(9), 15407-15407 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service