Skip to Content
Merck
All Photos(1)

Key Documents

RAB0555

Sigma-Aldrich

Human PLAU  / Urokinase-type Plasminogen Activator ELISA Kit

for serum, plasma, cell culture supernatants and urine

Synonym(s):

PLAU ELISA Kit, Urokinase Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type urine
sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 10 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PLAU (5328)

General description

The antibody pair provided in this kit detects Human uPA in serum, plasma, cell culture supernatants, and urine.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Ostheimer et al.
Strahlentherapie und Onkologie : Organ der Deutschen Rontgengesellschaft ... [et al], 194(6), 539-551 (2018-01-18)
The urokinase plasminogen activator system (uPA, uPAR, PAI‑1) is upregulated in cancer and high plasma levels are associated with poor prognosis. Their interaction with hypoxia-related osteopontin (OPN) which is also overexpressed in malignant tumors suggests potential clinical relevance. However, the
Tomasz W Kaminski et al.
International urology and nephrology, 50(1), 127-135 (2017-10-24)
Chronic kidney disease (CKD) is an estimated risk factor for increased mortality and morbidity due to fibrinolytic system disturbances. Progressive loss of renal function leads to retention of uremic toxins. Anthranilic acid (AA) is a tryptophan-derived uremic toxin with multidirectional
Hongwei Zhou et al.
Oncology letters, 14(5), 5189-5196 (2017-11-09)
The overexpression of the oncogene human epidermal growth factor receptor 2 (HER-2) has been associated with decreased disease-free survival and is a marker of poor prognosis of invasive breast cancer. Although the high efficacy of trastuzumab, a drug that targets
Marwa A M Sh El-Sharief et al.
Bioorganic chemistry, 80, 545-554 (2018-07-18)
Tetrahydroindolocarbazoles (THICZs) with versatile substituents, have been designed, synthesized, structure characterized, then investigated for their in-vitro anticancer screening, urokinase inhibition (uPA) evaluated, DNA-damage determination was further explored. Compounds 5, 8, 10 and 17 displayed the most promising antitumor activities against
Lutz Beckert et al.
JCI insight, 5 (2019-04-19)
Current dosing of intrapleural fibrinolytic therapy (IPFT) in adults with complicated parapneumonic effusion (CPE) / empyema is empiric, as dose-escalation trials have not previously been conducted. We hypothesized that LTI-01 (scuPA), which is relatively resistant to PA inhibitor-1 (PAI-1), would

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service