Skip to Content
Merck
All Photos(2)

Key Documents

RAB0433

Sigma-Aldrich

Human Siglec-5 ELISA Kit

for serum, plasma, cell culture supernatants and urine

Synonym(s):

OB-BP2, Obesity-binding protein 2, Sialic acid-binding Ig-like lectin 5, Siglec-5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

Quality Level

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 8 pg/mL
standard curve range: 6.86-5000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SIGLEC5(8778)

General description

The Human Siglec-5 (CD170) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human Siglec-5 and human Siglec-14 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human Siglec5

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinran Li et al.
Journal of immunology research, 2020, 3582648-3582648 (2020-05-01)
Premature ovarian insufficiency (POI) represents the hypergonadotropic hypoestrogenic symptoms that result in the loss of ovarian follicles. 5-30% POI cases are suggested to be involved in autoimmune etiology. MicroRNA-21 (miR-21) plays a vital role in ovarian folliculogenesis via regulating and
Meghan Angley et al.
Lupus science & medicine, 7(1) (2020-11-03)
Women with SLE may experience ovarian insufficiency or dysfunction due to treatment or disease effects. Anti-Müllerian hormone (AMH), a marker of ovarian reserve, has been examined in small populations of women with SLE with conflicting results. To date, these studies
Effect of radiotherapy for rectal cancer on female sexual function: a prospective cohort study.
A Svanström Röjvall et al.
The British journal of surgery, 107(5), 525-536 (2019-12-04)
P Melo et al.
Journal of assisted reproduction and genetics, 37(4), 855-863 (2020-02-08)
To investigate the impact of a 3-month course of intracortical injections of autologous platelet-rich plasma (PRP) upon ovarian reserve markers versus no intervention in women with low ovarian reserve prior to undergoing assisted reproductive technology (ART). Prospective controlled, non-randomized comparative
Stavroula Lila Kastora et al.
Journal of assisted reproduction and genetics, 37(7), 1623-1635 (2020-05-21)
Combination of transcriptomic and retrospective clinical data, to assess anti-Mullerian hormone (AMH) functionality at a cumulus cell level and evaluate AMH potential as a suitable marker for IVF outcomes (oocytes retrieved, number of day 3 embryos, gestation outcomes). Raw RNA-sequencing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service