Skip to Content
Merck
All Photos(2)

Key Documents

HPA012556

Sigma-Aldrich

Anti-SIGLEC8 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD329 antigen, Anti-CDw329, Anti-SAF-2, Anti-Sialic acid-binding Ig-like lectin 8 precursor, Anti-Sialoadhesin family member 2, Anti-Siglec-8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200
western blot: 0.04-0.4 μg/mL

immunogen sequence

RSCRKKSARPAAGVGDTGMEDAKAIRGSASQGPLTESWKDGNPLKKPPPAVAPSSGEEGELHYATLSFHKVKPQDPQGQEATDSEYSEIKIHKRETAETQACLRNHNPSSKEVRG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SIGLEC8(27181)

General description

SIGLEC8 (Sialic acid binding Ig-like lectin 8) encodes a 54kDa type 1 transmembrane protein belonging to the immunoglobulin superfamily (IgSF). It consists of a transmembrane region, a V-set Ig like domain at the N-terminal end and a set of downstream C2-set domains. It is expressed in the eosinophilic granulocytes, mast cells, and basophills.

Immunogen

Sialic acid-binding Ig-like lectin 8 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SIGLEC8 (Sialic acid binding Ig-like lectin 8) plays an important role in the eosinophil biology. It acts as an eosinophil-specific transmembrane receptor for sialic acid dependent binding. It has been reported that SIGLEC8 may be involved in the identification of and modulation of allergic cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71733

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K K Kikly et al.
The Journal of allergy and clinical immunology, 105(6 Pt 1), 1093-1100 (2000-06-16)
Eosinophils, basophils, and mast cells are believed to be the central tenet cells in allergic conditions including allergic rhinitis, asthma, and eczema. The molecular mechanisms underlying the recruitment of these cells to sites of allergic inflammation are poorly understood. Our
Bradford A Youngblood et al.
Cells, 10(1) (2020-12-31)
Siglecs (sialic acid-binding immunoglobulin-like lectins) are single-pass cell surface receptors that have inhibitory activities on immune cells. Among these, Siglec-8 is a CD33-related family member selectively expressed on human mast cells and eosinophils, and at low levels on basophils. These
G Foussias et al.
Biochemical and biophysical research communications, 278(3), 775-781 (2000-11-30)
Through efforts to investigate the CD33-like subgroup of sialic acid binding immunoglobulin-like lectins (Siglecs), which are believed to be located on chromosome 19q13.4, we have identified the precise genomic region containing the Siglec8 gene. It is located on chromosome 19q13.4
H Floyd et al.
The Journal of biological chemistry, 275(2), 861-866 (2000-01-08)
We describe the characterization of siglec-8, a novel sialic acid-binding immunoglobulin-like lectin that is expressed specifically by eosinophils. A full-length cDNA encoding siglec-8 was isolated from a human eosinophil cDNA library. Siglec-8 is predicted to contain three extracellular immunoglobulin-like domains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service