RAB0367
Human MMP-3 ELISA Kit
for serum, plasma, cell culture supernatant and urine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type urine
sample type serum
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 300 pg/mL
standard curve range: 0.41-100 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... MMP3(185250)
General description
The Human MMP-3 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human MMP-3 pro and active forms in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human MMP3
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Faxin Li et al.
Inflammation, 37(4), 1134-1141 (2014-02-18)
Rheumatoid arthritis (RA) is a chronic autoimmune disease with high morbidity and mortality. Fibroblast-like synoviocytes (FLS) in the synovial tissues play critical roles in joint destruction. Recent studies implicate the sumoylation in the regulation of the inflammation and arthritis. Thus
Xiaoqiang Su et al.
Journal of cellular biochemistry, 120(8), 13302-13309 (2019-03-21)
Inflammation has been demonstrated to be the key factor for intervertebral disc degeneration (IVD), which remains a major public health problem. Isofraxidin is a coumarin compound that possesses strong anti-inflammatory activity. However, the role of isofraxidin in IVD remains unclear.
Xue-Xuan Feng et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 63, 113-123 (2014-07-19)
Ultraviolet (UV) irradiation, known to generate reactive oxygen species (ROS) excessively and elicit inflammatory response, is a potent inducer for skin photoaging. Overproduction of ROS in conjunction with the resulting inflammation stimulate the over-expression of matrix metalloproteinases (MMPs), which in
Hadrian Platzer et al.
Journal of clinical medicine, 9(5) (2020-05-02)
Progressive loss of joint function in osteoarthritis (OA) is driven by degenerative and inflammatory processes and their complex interaction. Decoding the link between degeneration and inflammation is one of the most exciting approaches in understanding OA pathophysiology and holds the
C Andersson et al.
Clinical and experimental immunology, 177(1), 219-233 (2014-03-29)
Preclinical evidence supports targeting the C5a receptor (C5aR) in rheumatoid arthritis (RA). To support ongoing clinical development of an anti-C5aR monoclonal antibody, we have investigated for the first time the mechanism of action and the pharmacodynamics of a blocking anti-murine
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service