Skip to Content
Merck
All Photos(2)

Key Documents

RAB0104

Sigma-Aldrich

Mouse G-CSF / CSF3 ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.5 pg/mL
standard curve range: 0.62-150 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Csf3(12985)

General description

The Mouse G-CSF ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse G-CSF in serum, plasma, and cell culture supernatants.

Immunogen

Recombinant Mouse GCSF

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hematopoietic colony-stimulating factors: new players in tumor-nerve interactions.
Stosser S, Schweizerhof M, Kuner R.
Journal of Molecular Medicine, 89(4), 321-329 (2011)
Impaired production and increased apoptosis of neutrophils in granulocyte colony-stimulating factor receptor-deficient mice.
Liu F
Immunity, 5(5), 491-501 (1996)
Activity-dependent phosphorylation of Akt/PKB in adult DRG neurons.
Pezet S
The European Journal of Neuroscience, 21(7), 1785-1797 (2005)
Nociceptive-specific activation of ERK in spinal neurons contributes to pain hypersensitivity.
Ji RR
Nature Neuroscience, 2(12), 1114-1119 (1999)
The multifaceted effects of granulocyte colony-stimulating factor in immunomodulation and potential roles in intestinal immune homeostasis.
Martins A, Han J, Kim SO.
IUBMB Life, 62(8), 611-617 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service