Skip to Content
Merck
All Photos(2)

Key Documents

RAB0064

Sigma-Aldrich

Human MDC / CCL22 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type urine
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.5 pg/mL
standard curve range: 1.65-400 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL22(6367)

General description

Human MDC / CCL22 enzyme-linked immunosorbent assay (ELISA) kit is an in vitro assay for the quantitative measurement of macrophage-derived chemokine (MDC) protein in biological samples, such as serum, plasma, cell culture supernatants, urine, and/or cell and tissue lysates.

Immunogen

Recombinant Human MDC

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Macrophage-derived chemokine (MDC) signals through the C-C chemokine receptor type 4 (CCR4). It chemo-attracts monocytes, dendritic cells and natural killer (NK) cells and aids in their function. The protein exerts human immunodeficiency virus (HIV) suppressive activity. The 67 amino acid form of MDC displays reduced chemoattractant activity but retains HIV suppressive activity. It is expressed in various cancers and is involved in recruitment of regulatory T cells (Treg) within a tumor.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human macrophage-derived chemokine (MDC), a novel chemoattractant for monocytes, monocyte-derived dendritic cells, and natural killer cells.
Godiska R
The Journal of Experimental Medicine (1997)
Genetic Polymorphisms of CCL22 and CCR4 in Patients with Lung Cancer.
Erfani N
Iranian Journal of Medical Sciences (2014)
Suppression of intratumoral CCL22 by type i interferon inhibits migration of regulatory T cells and blocks cancer progression.
Anz D
Cancer Research (2015)
Soluble factors from T cells inhibiting X4 strains of HIV are a mixture of ? chemokines and RNases.
Cocchi F
Proceedings of the National Academy of Sciences of the USA (2012)
Yang Zhang et al.
Bioscience, biotechnology, and biochemistry, 84(6), 1191-1200 (2020-03-07)
The molecular mechanism of autophagy in Lactoferrin (LF) induced osteoblast differentiation is not fully demonstrated. In this study, alkaline phosphatase (ALP) activity, alizarin red S staining and ELISA were used to study N-terminal propeptide of type I procollagen (PINP) expression.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service