Skip to Content
Merck
All Photos(2)

Key Documents

R2523

Sigma-Aldrich

REDTaq® ReadyMix PCR Reaction Mix

Complete PCR reagent with standard Taq DNA Polymerase and inert dye

Synonym(s):

PCR master mix, PCR master mix with loading dye, Taq PCR master mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

Quality Level

form

liquid

usage

sufficient for 100 reactions
sufficient for 20 reactions

feature

dNTPs included
hotstart: no

technique(s)

PCR: suitable

color

red

input

purified DNA

application(s)

agriculture

shipped in

wet ice

storage temp.

−20°C

General description

REDTaq® ReadyMix is a ready-to-use mixture of Taq DNA polymerase, 99% pure DNTPs, reaction buffer, and an inert red dye in a 2X concentrate. After the PCR reaction, the PCR product can be loaded directly onto an agarose gel. The red loading dye migrates at approximately the same rate as a 125 base pair fragment in a 1% agarose gel. The inert dye has no effect on the amplification process, and therefore, a sample can be easily re-amplified such as in “nested PCR”.

Application

REDTaq® ReadyMix PCR Reaction Mix has been used:
  • for routine polymerase chain reaction (PCR) amplification.
  • for the PCR amplification of cDNA
  • in semi-quantitative PCR

Features and Benefits

The benefits of REDTaq® with the convenience of ReadyMix
  • Saves preparation time
  • Reduces risk of contamination from multiple pipet steps and and increased reproducibility.
  • Provides consistent reaction-to-reaction performance
  • No loading buffer/tracking dye needs to be added. The PCR product is loaded directly into an agarose gel after amplification
  • The red tracer migrates at approximately the same rate as a 125bp fragment (slightly faster than bromophenol blue)
  • Amplifies targets up to 7 kb in length
  • Because no loading buffer has been added, a sample can be easily re-amplifed such as in nested PCR.
  • Stable at 4°C or −20°C.
  • Ready-mix format reduces setup time.

Packaging

Default reaction volume is 50 μL

20RXN is packaged as 1 X 500 μL
100RXN is packaged as 1 X 2.5 mL

Preparation Note

Add 25 μl of ReadyMix to template, primers and water to a final reaction volume of 50 μl. The PCR product is easily separated from the dye by standard purification methodologies. The inert red dye has no effect on automated sequencing and is suitable for restriction enzyme digestion, ligation and other downstream manipulations.

Legal Information

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
REDTaq is a registered trademark of Merck KGaA, Darmstadt, Germany
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RNA activation of the vascular endothelial growth factor gene (VEGF) promoter by double-stranded RNA and hypoxia: role of noncoding VEGF promoter transcripts
Lopez P, et al.
Molecular and Cellular Biology, 36(10), 1480-1493 (2016)
Transforming growth factor-beta-inducible early response gene 1 is a novel substrate for atypical protein kinase Cs
Alemu EA, et al.
Cellular and Molecular Life Sciences, 68(11), 1953-1968 (2011)
Tamar Guy-Haim et al.
PeerJ, 6, e5268-e5268 (2018-07-31)
Aquatic subterranean species often exhibit disjunct distributions, with high level of endemism and small range, shaped by vicariance, limited dispersal, and evolutionary rates. We studied the disjunct biogeographic patterns of an endangered blind cave shrimp, Typhlocaris, and identified the geological
Alyssa A Hoover et al.
BMC cancer, 20(1), 970-970 (2020-10-09)
New treatment options for ovarian cancer are urgently required. Tumor-associated macrophages (TAMs) are an attractive target for therapy; repolarizing TAMs from M2 (pro-tumor) to M1 (anti-tumor) phenotypes represents an important therapeutic goal. We have previously shown that upregulated NF-kappaB (NF-κB)
Maya Miller et al.
iScience, 23(7), 101262-101262 (2020-06-26)
PhenolaTi is an advanced non-toxic anticancer chemotherapy; this inert bis(phenolato)bis(alkoxo) Ti(IV) complex demonstrates the intriguing combination of high and wide efficacy with no detected toxicity in animals. Here we unravel the cellular pathways involved in its mechanism of action by

Articles

Colony PCR reagents and our colony PCR protocol using REDExtract-N-Amp™ PCR ReadyMix™ and JumpStart™ REDTaq® PCR ReadyMix™ reagents.

Explore PCR's history, from discovery to Nobel Prize. Discover real-time PCR (qPCR) and digital PCR developments.

Explore PCR's history, from discovery to Nobel Prize. Discover real-time PCR (qPCR) and digital PCR developments.

Explore PCR's history, from discovery to Nobel Prize. Discover real-time PCR (qPCR) and digital PCR developments.

See All

Protocols

Reviews the applications and benefits for RedTaq, including standard RedTaq, Hot Start RedTaq and RedTaq for genomic DNA PCR.

Learn standard PCR protocol steps and review reagent lists or cycling parameters. This method for routine PCR amplification of DNA uses standard Taq DNA polymerase.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service