Skip to Content
Merck
All Photos(2)

Key Documents

P4600

Sigma-Aldrich

ReadyMix Taq PCR Reaction Mix

with MgCl2

Synonym(s):

PCR master mix, Taq PCR master mix

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106300
NACRES:
NA.55

Quality Level

form

liquid

usage

sufficient for 100 reactions

feature

dNTPs included
hotstart: no

concentration

1.5 units/reaction (50 μL reaction volume)

technique(s)

PCR: suitable

color

colorless

input

purified DNA

application(s)

agriculture

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

ReadyMix Taq PCR Reaction Mix is a prepared solution containing everything needed for a PCR reaction except the specific primers and template. The mix includes Sigma′s high quality Taq DNA Polymerase, 99% pure deoxynucleotides and buffer in a 2× optimized reaction concentrate. For reaction set-up, add the ReadyMix (25 μL) to the primers, template and water (total volume 50 μL). Using ReadyMix Taq PCR Reaction Mix reduces pipetting steps and risk of contamination. This saves time and reduces errors while still providing the great performance of Sigma′s Taq DNA Polymerase.

Application

ReadyMix Taq PCR Reaction Mix has been used:
  • in the development and specificity test of the amplified fragment length polymorphism (AFLP) markers
  • in the PCR reaction for terminal restriction fragment length polymorphism (T-RFLP) analysis
  • in the molecular identification of yeasts
  • for routine PCR amplifications

Features and Benefits

  • Amplifies targets up to 7 kb in length

  • ReadyMix Taq PCR Reaction Mix offers the same great performance as Taq DNA Polymerase in a more convenient formulation

  • ReadyMix format reduces setup time.

  • Reduced risk of contamination from multiple pipet steps.

Packaging

Default reaction volume is 50 μL

100RXN is packaged as 1 X 2.5 mL
Supplied with a vial of PCR grade water for dilution.

Unit Definition

One unit incorporates 10 nmol of total dNTPs into acid-precipitable DNA in 30 min. at 74 °C.

Legal Information

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assessment of fecal microflora changes in pigs supplemented with herbal residue and prebiotic
Samanta A K, et al.
PLoS ONE, 10(7), e0132961-e0132961 (2015)
Establishment of an in vitro trans-splicing system in Trypanosoma brucei that requires endogenous spliced leader RNA
Shaked H, Wachtel C, Tulinski P, Yahia NH et al.
Nucleic Acids Research, 38, 114-114 (2010)
A Dimitroglou et al.
Journal of animal science, 87(10), 3226-3234 (2009-07-21)
A study was conducted to investigate the effect of mannan oligosaccharide (MOS) on the gut microbiota and intestinal morphology of rainbow trout under commercial farming conditions. Juvenile (mean initial BW 38.2 +/- 1.7 g) and subadult (111.7 +/- 11.6 g)
Chitosan inactivates spoilage yeasts but enhances survival of Escherichia coli O157: H7 in apple juice
Kisko G, et al.
Journal of Applied Microbiology, 98(4), 872-880 (2005)
Ashis Kumar Samanta et al.
PloS one, 10(7), e0132961-e0132961 (2015-07-16)
Antibiotic usage in animals as a growth promoter is considered as public health issue due to its negative impact on consumer health and environment. The present study aimed to evaluate effectiveness of herbal residue (ginger, Zingiber officinale, dried rhizome powder)

Articles

Explore PCR's history, from discovery to Nobel Prize. Discover real-time PCR (qPCR) and digital PCR developments.

Explore PCR's history, from discovery to Nobel Prize. Discover real-time PCR (qPCR) and digital PCR developments.

Explore PCR's history, from discovery to Nobel Prize. Discover real-time PCR (qPCR) and digital PCR developments.

Explore PCR's history, from discovery to Nobel Prize. Discover real-time PCR (qPCR) and digital PCR developments.

See All

Protocols

Learn standard PCR protocol steps and review reagent lists or cycling parameters. This method for routine PCR amplification of DNA uses standard Taq DNA polymerase.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service