Skip to Content
Merck
All Photos(3)

Key Documents

145920

Sigma-Aldrich

Quinine monohydrochloride dihydrate

90%

Synonym(s):

Quinine hydrochloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O2 · HCl · 2H2O
CAS Number:
Molecular Weight:
396.91
Beilstein:
6112655
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

90%

form

solid

optical activity

[α]20/D −144°, c = 1 in H2O

mp

115-116 °C (dec.) (lit.)

SMILES string

Cl[H].[H]O[H].[H]O[H].[H][C@@]12CCN(C[C@@H]1C=C)[C@@]([H])(C2)[C@H](O)c3ccnc4ccc(OC)cc34

InChI

1S/C20H24N2O2.ClH.2H2O/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;;;/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;1H;2*1H2/t13-,14-,19-,20+;;;/m0.../s1

InChI key

MPQKYZPYCSTMEI-FLZPLBAKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quinine monohydrochloride dihydrate is commonly used as a standard for bitterness in sensory evaluation experiments of food samples.

Other Notes

Remainder hydroquinine hydrochloride dihydrate

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prediction of grape polyphenol astringency by means of a fluorimetric micro-plate assay
Fia G, et al.
Food Chemistry, 113(1), 325-330 (2009)
Benjamin Troutwine et al.
Alcoholism, clinical and experimental research, 43(12), 2480-2493 (2019-10-09)
N-methyl-D-aspartate (NMDA) receptors regulate synaptic plasticity and modulate a wide variety of behaviors. Mammalian NMDA receptors are inhibited by ethanol (EtOH) even at low concentrations. In mice, the F639A mutation in transmembrane domain (TMD) 3 of the NR1 subunit reduces
Temporary modification of salivary protein profile and individual responses to repeated phenolic astringent stimuli.
Dinnella C, et al.
Chemical Senses, 35(1), 75-85 (2010)
Hao Jin et al.
Cell, 184(1), 257-271 (2021-01-09)
Hardwired circuits encoding innate responses have emerged as an essential feature of the mammalian brain. Sweet and bitter evoke opposing predetermined behaviors. Sweet drives appetitive responses and consumption of energy-rich food sources, whereas bitter prevents ingestion of toxic chemicals. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service