Skip to Content
Merck
All Photos(2)

Key Documents

PZ0021

Sigma-Aldrich

Tigecycline hydrate

≥98% (HPLC)

Synonym(s):

9-t-Butylglycylamido-minocycline hydrate, TBG-MINO, Tigecyclin, Tygacil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H39N5O8 · xH2O
CAS Number:
Molecular Weight:
585.65 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -175 to -205°, c = 0.15 in methanol

color

off-white to yellow-brown

solubility

DMSO: ≥3 mg/mL (warmed)

storage temp.

2-8°C

SMILES string

O.CN(C)[C@H]1[C@@H]2C[C@@H]3Cc4c(cc(NC(=O)CNC(C)(C)C)c(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)N(C)C

InChI

1S/C29H39N5O8.H2O/c1-28(2,3)31-11-17(35)32-15-10-16(33(4)5)13-8-12-9-14-21(34(6)7)24(38)20(27(30)41)26(40)29(14,42)25(39)18(12)23(37)19(13)22(15)36;/h10,12,14,21,31,36,38-39,42H,8-9,11H2,1-7H3,(H2,30,41)(H,32,35);1H2/t12-,14-,21-,29-;/m0./s1

InChI key

PMPLAEFBZIFHAR-KXLOKULZSA-N

General description

Tigecycline is used to treat complicated skin and skin structure infections (cSSSI) and complicated intraabdominal infections.

Application

TIGECYCLINE HYDRATE has been used:
  • for antibiotic susceptibility testing
  • to study its effects on multiple nonsmall cell lung cancer (NSCLC) cell lines
  • to evaluate its effects on the binding of fluoroquinolones to human serum albumin
  • as a standard to investigate the penetration of tigecycline into cornea, aqueous humor and vitreous humor

Biochem/physiol Actions

Tigecycline is a broad spectrum glycylcycline antibiotic.
Tigecycline is a broad spectrum glycylcycline antibiotic. Tigecycline is bacteriostatic, that inhibits protein synthesis by binding to the 30S ribosomal subunit of bacteria and thereby blocking entry of Aminoacyl-tRNA into the A site of the ribosome during prokaryotic translation. Tigecycline is active against resistant strains of gram-positive and gram-negative bacteria, avoiding the two most common mechanisms of resistance to the tetracycline antimicrobials: tetracycline efflux pump proteins and ribosomal protection proteins.

Other Notes

air sensitive

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasar Sakarya et al.
International journal of ophthalmology, 10(5), 679-683 (2017-05-27)
To evaluate ocular penetration of topically applied 1% tigecycline. Forty-two New Zealand White rabbits were divided into 3 groups. A 50 µL drop of 1% tigecycline was administered in group 1. In groups 2 and 3, the drop was administered
Tigecycline targets nonsmall cell lung cancer through inhibition of mitochondrial function
Jia X, et al.
Fundamental & Clinical Pharmacology, 30(4), 297-306 (2016)
A Jasiecka-Mikołajczyk et al.
Journal of veterinary pharmacology and therapeutics, 41(1), e22-e29 (2017-08-24)
The aim of this research had been to determine the pharmacokinetics of tigecycline (TIG) in turkey after intravenous (i.v.), intramuscular (i.m.), subcutaneous (s.c.), and oral (p.o.) administration at a dose of 10 mg/kg. TIG concentrations in plasma were determined using high-performance
The Effect of Tigecycline on the Binding of Fluoroquinolones to Human Serum Albumin
Jelic RM, et al.
Serbian Journal of Experimental and Clinical Research, 19(1), 17-25 (2018)
Hubert Ziółkowski et al.
Poultry science, 99(10), 4750-4757 (2020-09-30)
Tetracyclines continue to be important antimicrobials in veterinary medicine. However, the pharmacokinetics (PK) of tigecycline (TIG) and minocycline (MIN) in broiler chickens has not been investigated to date, and the PK of chlortetracycline (CTC) and tetracycline (TET) remains insufficiently researched

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service