Skip to Content
Merck
All Photos(3)

Key Documents

A9789

Sigma-Aldrich

Anisomycin

from Streptomyces griseolus, ≥98% (HPLC), solid, protein synthesis inhibitor

Synonym(s):

(2R,3S,4S)-2-(4-Methoxybenzyl)-3,4-pyrrolidinediol-3-acetate, 2-[(4-Methoxyphenyl)methyl]-3,4-pyrrolidinediol 3-acetate, Flagecidin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO4
CAS Number:
Molecular Weight:
265.30
Beilstein:
20705
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Anisomycin from Streptomyces griseolus, ≥98% (HPLC), solid

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white to faint yellow

solubility

H2O: 2 mg/mL (Aqueous solutions are stable over a wide range of pH at room temperature.)
DMSO: 20 mg/mL
methanol: 20 mg/mL

antibiotic activity spectrum

neoplastics

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

COc1ccc(C[C@H]2NC[C@H](O)[C@H]2OC(C)=O)cc1

InChI

1S/C14H19NO4/c1-9(16)19-14-12(15-8-13(14)17)7-10-3-5-11(18-2)6-4-10/h3-6,12-15,17H,7-8H2,1-2H3/t12-,13+,14+/m1/s1

InChI key

YKJYKKNCCRKFSL-RDBSUJKOSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Antibiotic isolated from Streptomyces griseolus that inhibits protein synthesis. Acts by inhibiting peptidyl transferase activity in eukaryote ribosomes. Reported to induce apoptosis in a variety of cells including promyelocytic leukemia cells, Jurkat cells, ventricular myocytes, and colon adenocarcinoma cells. Initiates intracellular signals and immediate early gene induction. Selective signaling agonist. Potent Jun-NH2 terminal kinase (JNK) agonist. Activates mitogen-activated protein (MAP) kinases (JNK/SAPK and p38/RK). Antiprotozoal agent.

Caution

Hygroscopic

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Puromycin reactivity does not accurately localize translation at the subcellular level
Enam , Syed Usman et al.
eLife, 9, e60303-e60303 (2020)
Verónica de la Fuente et al.
Scientific reports, 9(1), 12157-12157 (2019-08-23)
Memories are a product of the concerted activity of many brain areas. Deregulation of consolidation and reprocessing of mnemonic traces that encode fearful experiences might result in fear-related psychopathologies. Here, we assessed how pre-established memories change with experience, particularly the
J Daubriac et al.
Cell death and differentiation, 16(8), 1146-1155 (2009-04-04)
Pleural fluid accumulation is a frequent clinical observation in diffuse malignant pleural mesothelioma (MPM). The cytological analysis of pleural fluid often reveals the presence of free spheroid aggregates of malignant cells, giving rise to the question of the ability of
Stéphane Delmas et al.
Molecular microbiology, 87(1), 168-179 (2012-11-14)
In prokaryotes the genome is organized in a dynamic structure called the nucleoid, which is embedded in the cytoplasm. We show here that in the archaeon Haloferax volcanii, compaction and reorganization of the nucleoid is induced by stresses that damage
Einar Ö Einarsson et al.
Learning & memory (Cold Spring Harbor, N.Y.), 19(10), 449-452 (2012-09-18)
It has been suggested that memories become more stable and less susceptible to the disruption of reconsolidation over weeks after learning. Here, we test this by targeting the anterior cingulate cortex (ACC) and test its involvement in the formation, consolidation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service