Skip to Content
Merck

PECAS9

Sigma-Aldrich

PURedit® Cas9 Protein

from Streptococcus pyogenes, recombinant, expressed in E. coli, 3X NLS

Synonym(s):

Cas9, SpCas9, SpyCas9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.51

Pricing and availability is not currently available.

recombinant

expressed in E. coli

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

packaging

pkg of 1 kit (4 components)

shipped in

wet ice

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
CAS9PLESPCAS9PRODCAS9PROT
recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

shipped in

wet ice

shipped in

wet ice

shipped in

ambient

shipped in

ambient

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

300

Quality Level

200

Quality Level

300

Quality Level

200

General description

Recombinant PURedit Cas9 protein from Streptococcus pyogenes (~160 KD) is a ready-to-use reagent for genome engineering experiments. When combined with target-specific guide RNAs, Streptococcus pyogenes PURedit Cas9 protein will act as a targeted nuclease suitable for transfection of cell cultures and for the accelerated development of genetically-modified animals via one-cell embryo injection.

PURedit translational-grade CRISPR reagents are engineered to meet superior manufacturing standards that enable consistent, high-quality performance every time.

Application

Functional Genomics; Target Validation; Genome Editing

Features and Benefits

  • High Quality for preclinical and early clinical research
  • Highest performance efficiency of any Cas9 variant
  • Ready to inject/transfect

Packaging

pkg of 50 μg (≥ 300 pmol)
pkg of 250 μg (≥ 1500 pmol)

Components

Each kit consists of:
  • one vial of PURedit Cas9 Recombinant Protein
  • one vial of PURedit Transfection Buffer
  • one vial of Reconstitution Buffer
  • one vial of Dilution Buffer

Principle

CRISPR/Cas systems are employed by bacteria and archaea as a defense against invading viruses and plasmids. Recently, the type II CRISPR/Cas system from the bacterium Streptococcus pyogenes has been engineered to function in eukaryotic systems using two molecular components: a single Cas9 protein and a non-coding guide RNA (gRNA). The Cas9 endonuclease can be programmed with a gRNA, directing a DNA double-strand break (DSB) at a desired genomic location. Similar to DSBs induced by zinc finger nucleases (ZFNs), the cell then activates endogenous DNA repair processes, either non-homologous end joining (NHEJ) or homology-directed repair (HDR), to heal the targeted DSB.

Reconstitution

Lyophilized PURedit Cas9 protein should be resuspended in the Reconstitution solution provided to desired concentration. Gently tap tube to completely dissolve lyophilized powder, incubate for 10 minutes on ice, and spin tube to bring material to bottom of tube.

Other Notes

Use our CRISPR Selection Tool to order gRNA

Check out our other MISSION® Cas9 Proteins at SigmaAldrich.com/CRISPRproteins

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
PURedit is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Guaranteed PURedit™ CRISPR synthetic gRNAs and Cas9 protein offer industry-leading on-site cutting and specificity

Guaranteed PURedit™ CRISPR synthetic gRNAs and Cas9 protein offer industry-leading on-site cutting and specificity

Guaranteed PURedit™ CRISPR synthetic gRNAs and Cas9 protein offer industry-leading on-site cutting and specificity

Guaranteed PURedit™ CRISPR synthetic gRNAs and Cas9 protein offer industry-leading on-site cutting and specificity

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service