P8616
Nunc® MicroWell™ 96 well polystyrene plates
Nunclon® Delta, white, well volume 400 μL, case of 50 ea
Synonym(s):
96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates, ELISA plates, fluorescence-based cell assay plates, immunoassay plates
About This Item
Recommended Products
material
flat bottom white polystyrene wells
polystyrene
white
sterility
sterile
feature
lid
pinchbar design
packaging
case of 50 ea
manufacturer/tradename
Nunc 136101
plate L × W
128 mm × 86 mm
size
96 wells
well volume
400 μL
working volume
250 μL
suitability
suitable for (fluorescence-based cell assays)
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | P8741 | TMO136102 | TMO137103 |
---|---|---|---|
flat bottom white polystyrene wells | black , flat bottom black polystyrene wells | polystyrene , white | black , polystyrene |
Tissue Culture (TC)-treated surface | Tissue Culture (TC)-treated surface | - | - |
sterile | sterile | sterile | sterile |
Nunc 136101 | Nunc 137101 | Nunc 136102 | Nunc 137103 |
suitable for (fluorescence-based cell assays) | suitable for (fluorescence-based cell assays) | suitable for (fluorescence-based cell assays) | suitable for (fluorescence-based cell assays) |
General description
- White plates provide maximum reflection, minimum auto-fluorescence and autoluminescence
- Black plates provide minimal background and light scatter in fluorescence assays
- Non-toxic white or black plate is provided with clear lid
- Raised well rims of flat bottom wells reduce risk of cross contamination
- Low crosstalk
- Nunclon® Δ for cell based assays
- Working volume range: 50-250 μl/well
- Suitable for fluorescence-based cell assays
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service