P6991
Nunc® MicroWell™ 96 well polypropylene plates
96 well plate, polypropylene, natural, round bottom, 500uL/well, sterile, 120/cs
Synonym(s):
96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate
About This Item
natural polypropylene plate
polypropylene
high flange design
Recommended Products
material
conical bottom natural polypropylene wells
natural polypropylene plate
polypropylene
sterility
sterile
feature
lid: no
high flange design
packaging
case of 120 ea (internal packs of 10)
manufacturer/tradename
Nunc 267334
plate L × W
128 mm × 86 mm
size
96 wells
well maximum volume
450 μm
working volume
500 μL
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Compatible with robotics and automated systems
- Polypropylene resin is resistant to most chemicals, solvents and alcohols
- Minimal binding of molecules with hydrophilic moities
- Non-toxic
- Products come without lids
- 0.5 ml volume in standard MicroWell™ format
- Shared wall technology increases well volume and wider cells improve mixing
- Low binding polypropylene for homogeneous assays and storage
- Ideal as collection plate for Nunc Filter Plates
- White plates for fluorescence and luminescence applications
- Black plates for fluorescence applications
- Yellow, red, natural and blue plates for storage applications
- Resistant to most solvents used in combinatorial chemistry
- Working volume range: 20 - 450 μl/well
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service