Skip to Content
Merck
All Photos(1)

Key Documents

P7998

Sigma-Aldrich

Alkaline Phosphatase Yellow

alkaline phosphatase substrate, chromogenic liquid

Synonym(s):

Alkaline phosphatase substrate solution (pNPP)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
eCl@ss:
42010105
NACRES:
NA.83

product name

Alkaline Phosphatase Yellow (pNPP) Liquid Substrate System for ELISA, ready to use solution

Quality Level

form

liquid

storage temp.

2-8°C

General description

The product is ready to use buffered alkaline phosphatase substrate consist of p-nitrophenylphosphate (pNPP). This is the advanced substrate system contains insoluble stabilizer that enhances the stability and reactivity as well as reduces the base-line absorption. It produces a soluble end product, hence not recommended for blotting or histochemistry applications.

Application

Develops a soluble yellow reaction product that may be read at 405 nm. For endpoint assays, the reaction may be stopped with 3 N NaOH. Recommended for ELISA (microwell) procedures; not recommended for membrane applications.

Caution

 Stable at least one year at 2-8 °C.

Physical form

Ready to use

Preparation Note

Contains p-nitrophenyl phosphate, a substrate for alkaline phosphatase.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura C Jacques et al.
Nature communications, 11(1), 1892-1892 (2020-04-22)
Streptococcus pneumoniae serotype 1 is the predominant cause of invasive pneumococcal disease in sub-Saharan Africa, but the mechanism behind its increased invasiveness is not well understood. Here, we use mouse models of lung infection to identify virulence factors associated with
Cristina A Teixeira et al.
Clinical science (London, England : 1979), 130(18), 1665-1675 (2016-07-08)
Transthyretin (TTR)-related amyloidoses are diseases characterized by extracellular deposition of amyloid fibrils and aggregates in tissues composed of insoluble misfolded TTR that becomes toxic. Previous studies have demonstrated the ability of small compounds in preventing and reversing TTR V30M deposition
Baoxin Wang et al.
Experimental and therapeutic medicine, 13(1), 178-186 (2017-01-27)
Damage to the recurrent laryngeal nerve often causes hoarseness, dyspnea, dysphagia, and sometimes asphyxia due to vocal cord paralysis which result in a reduction of quality of life. Brain-derived neurotrophic factor (BDNF) and glial cell line-derived neurotrophic factor (GDNF) play
Hannah M Rowe et al.
Infection and immunity, 87(10) (2019-07-17)
Acute otitis media is one of the most common childhood infections worldwide. Currently licensed vaccines against the common otopathogen Streptococcus pneumoniae target the bacterial capsular polysaccharide and confer no protection against nonencapsulated strains or capsular types outside vaccine coverage. Mucosal
Xin-Yu Guo et al.
The Journal of biological chemistry, 295(48), 16393-16410 (2020-09-25)
The protein folding and lipid moiety status of glycosylphosphatidylinositol-anchored proteins (GPI-APs) are monitored in the endoplasmic reticulum (ER), with calnexin playing dual roles in the maturation of GPI-APs. In the present study, we investigated the functions of calnexin in the

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service