Skip to Content
Merck
All Photos(1)

Key Documents

P7794

Sigma-Aldrich

Penicillin G potassium salt

powder, suitable for cell culture, BioReagent

Synonym(s):

Benzylpenicillin potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17KN2O4S
CAS Number:
Molecular Weight:
372.48
Beilstein:
3832841
EC Number:
MDL number:
UNSPSC Code:
12352207
eCl@ss:
34010400
PubChem Substance ID:
NACRES:
NA.76

product name

Penicillin G potassium salt, powder, BioReagent, suitable for cell culture

product line

BioReagent

Quality Level

form

powder

potency

1440-1680 units per mg

technique(s)

cell culture | mammalian: suitable

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

SMILES string

[K+].[H][C@]12SC(C)(C)[C@@H](N1C(=O)[C@H]2NC(=O)Cc3ccccc3)C([O-])=O

InChI

1S/C16H18N2O4S.K/c1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);/q;+1/p-1/t11-,12+,14-;/m1./s1

InChI key

IYNDLOXRXUOGIU-LQDWTQKMSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Application

Penicillin G is a narrow spectrum antibiotic derived from Streptococcus pneumoniae. It is the drug of choice for groups A, B, C and G streptococci, nonenterococcal group D streptococci, viridians group streptococci, and non-penicillinase producing staphylococcus. The potassium salt has been used to study murosomes of staphylococci and the penicillin-induced lysis of Streptococcus mutans.

Biochem/physiol Actions

Mode of Action: Penicillin G acts by inhibiting cell wall synthesis through binding to penicillin binding proteins (PBPs), inhibiting peptidoglycan chain cross-linking.

Antimicrobial spectrum: This product is active against gram-positive and gram-negative bacteria.

Caution

Solutions should be filter sterilized and stored at 2-8°C for 1 week or at -20°C for more lengthy periods. Solutions are stable at 37°C for 3 days. The potassium salt is soluble in H2O at 100 mg/mL.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yulei Zhao et al.
American journal of translational research, 13(6), 7077-7082 (2021-07-27)
To explore the CD200/CD200R expression level in the peripheral blood mononuclear cells (PBMC) of hematological malignancy patients and to analyze its mechanism of action. Thirty hematological malignancy patients who were hospitalized in our hospital from June 2019 to December 2019
R Comunian et al.
Journal of dairy science, 93(12), 5644-5650 (2010-11-26)
Antibiotics are widely used for therapeutic and prophylactic purposes in dairy animals. The presence of residual antibiotics in milk could cause potentially serious problems in human health and have technological implication in the manufacturing of dairy products. The aim of
Lavinia Florina Călinoiu et al.
Biomolecules, 10(1) (2019-12-28)
The aim of the present paper was to identify the major polyphenolic compounds and investigate the antioxidant, antimutagenic, and antimicrobial activities of industrially-derived cereal byproducts-wheat bran (WB) and oat bran (OB)-before (fresh) and after thermal processing (TP) (10 min, 80
Bianca-Eugenia Ștefănescu et al.
Plants (Basel, Switzerland), 9(9) (2020-09-17)
Blueberries have seen an ascending production line boosted by World Health Organization (WHO) approvals for their contributions to a healthy diet and the evidence that they act against different diseases. This increase resulted in significant amounts of discarded leaves, which
Sammy Detry et al.
Biochemical pharmacology, 165, 240-248 (2019-03-20)
The four core members of the Interleukin-12 (IL-12) family of cytokines, IL-12, IL-23, IL-27 and IL-35 are heterodimers which share α- and β-cytokine subunits. All four cytokines are immune modulators and have been proposed to play divergent roles in inflammatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service